Freeze-dried bone in pulpotomy procedures in monkey.
The Journal of pedodontics (1989-01-01)
S Fadavi, A W Anderson, I C Punwani
摘要
The purpose of this study was to evaluate the effect of freeze-dried bone on amputated pulps. Fifteen primary and one permanent monkey teeth were treated with freeze-dried bone. As the control group, another fifteen primary teeth were treated with calcium hydroxide and fifteen primary and one permanent teeth with formocresol. Four other primary teeth pulps were amputated and sealed with tin foil and IRM as controls. Twelve mandibular incisors were left intact. Histologically all but three teeth treated with human freeze-dried bone after three months showed a complete or partial calcific barrier directly below the amputation site. Normal appearing odontoblastic cells were noted below the calcific barrier. The apical third was vital with an occasional chronic inflammatory cell visible. The histological findings of teeth treated with calcium-hydroxide were very similar to freeze-dried bone. All but four teeth showed a complete calcific barrier at the amputation site. The odontoblastic cells were normal in appearance and the inflammatory cell reactions decreased from the middle portion toward the apical region. The histological evaluation of the formocresol-treated teeth was comparable with previously published studies.
材料
货号
品牌
产品描述
锡, foil, 150x150mm, thickness 0.20mm, as rolled, 98.8%
锡, foil, 0.5m coil, thickness 0.009mm, 97.4%
锡, foil, 100x100mm, thickness 0.125mm, as rolled, 98.8%
锡, foil, 100x100mm, thickness 0.125mm, as rolled, 99.99+%
锡, foil, 100x100mm, thickness 0.15mm, as rolled, 98.8%
锡, foil, 100x100mm, thickness 0.15mm, as rolled, 99.99+%
锡, foil, 100x100mm, thickness 0.1mm, as rolled, 98.8%
锡, foil, 100x100mm, thickness 0.1mm, as rolled, 99.95%
锡, foil, 100x100mm, thickness 0.20mm, as rolled, 98.8%
锡, foil, 100x100mm, thickness 0.25mm, as rolled, 98.8%
锡, foil, 100x100mm, thickness 0.25mm, as rolled, 99.99+%
锡, foil, 100x100mm, thickness 0.5mm, as rolled, 98.8%
锡, foil, 100x100mm, thickness 0.5mm, as rolled, 99.99+%
锡, foil, 100x100mm, thickness 0.65mm, as rolled, 99.99+%
锡, foil, 100x100mm, thickness 0.75mm, as rolled, 99.99+%
锡, foil, 100x100mm, thickness 1.0mm, as rolled, 98.8%
锡, foil, 100x100mm, thickness 1.0mm, as rolled, 99.99+%
锡, foil, 100x100mm, thickness 1.0mm, as rolled, 99.999%
锡, foil, 100x100mm, thickness 1.5mm, as rolled, 99.99+%
锡, foil, 100x100mm, thickness 2.0mm, as rolled, 98.8%
锡, foil, 100x100mm, thickness 2.0mm, as rolled, 99.99+%
锡, foil, 150x150mm, thickness 0.125mm, as rolled, 98.8%
锡, foil, 150x150mm, thickness 0.125mm, as rolled, 99.99+%
锡, foil, 150x150mm, thickness 0.15mm, as rolled, 98.8%
锡, foil, 150x150mm, thickness 0.1mm, as rolled, 98.8%
锡, foil, 150x150mm, thickness 0.1mm, as rolled, 99.95%
锡, foil, 150x150mm, thickness 0.25mm, as rolled, 98.8%
锡, foil, 150x150mm, thickness 0.25mm, as rolled, 99.99+%
锡, foil, 150x150mm, thickness 0.5mm, as rolled, 98.8%
锡, foil, 150x150mm, thickness 0.5mm, as rolled, 99.99+%
社交媒体
LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon
Merck
科研、开发、生产。
作为生命科学行业的全球领先供应商,我们致力于为科研、生物技术开发和生产,以及制药药物疗法开发和生产提供各类解决方案和服务。
© 2021年版权归德国达姆施塔特默克集团(Merck KGaA)及/或其附属公司所有。版权所有。
未经许可,严禁复制本网站上的任何资料。