  • Search Results
  • Vasoactive effects of prostaglandins from the perivascular fat of mesenteric resistance arteries in WKY and SHROB rats.
Vasoactive effects of prostaglandins from the perivascular fat of mesenteric resistance arteries in WKY and SHROB rats.
Life sciences (2013-11-10)
Yolanda Mendizábal, Silvia Llorens, Eduardo Nava
ABSTRACT
We have studied the vasoactive role of prostaglandins derived from perivascular adipose tissue (PVAT) and their effects on endothelial function in healthy rats and rats with metabolic syndrome (SHROB). Mesenteric resistance arteries (MRA) from SHROB and control rats (WKY) were mounted on wire myographs: a) together with a sphere of naturally occurring perivascular adipose tissue (with-PVAT group), or b) dissecting all the adventitial tissue (without-PVAT group). Endothelial function, tested by acetylcholine reactivity of SHROB arteries with PVAT, was significantly lower than that of WKY. With-PVAT arteries, especially the SHROB, showed lower responses than those without PVAT. NO synthase inhibition diminished the acetylcholine responses in every group except the with-PVAT SHROB group. Blockade of cyclooxygenase-2, PGI2-IP, TXA2-TP, or TXA2 synthase increased to different extents the arterial responses in the SHROB with-PVAT group. PVAT from both rat strains revealed cyclooxygenase-2 activity and immunoassay confirmed the release of PGE2, PGI2 and TXA2. Our major finding is that PVAT is a source of vasoactive prostaglandins in WKY and SHROB. We also report that the presence of visceral PVAT causes endothelial dysfunction of resistance arteries in the SHROB. The vascular responses to prostaglandins partly underlie the endothelial dysfunction of SHROB arteries.
Sigma-Aldrich
Nω-硝基-L-精氨酸甲酯 盐酸盐, ≥98% (TLC), powder
Sigma-Aldrich
氯化乙酰胆碱, ≥99% (TLC)
Sigma-Aldrich
前列腺素E2, synthetic, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
前列腺素E2, ≥93% (HPLC), synthetic
Sigma-Aldrich
前列腺素E2, γ-irradiated, powder, BioXtra, suitable for cell culture
Sigma-Aldrich
氯化乙酰胆碱, suitable for cell culture
Sigma-Aldrich
碘化乙酰胆碱, ≥97%
Sigma-Aldrich
氯化乙酰胆碱, pkg of 150 mg (per vial)
Sigma-Aldrich
乙酰胆碱高氯酸盐
前列腺素E2, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
溴化乙酰胆碱(拟胆碱药), ≥99%
Sigma-Aldrich
氯化乙酰胆碱, ≥99% (TLC), free-flowing, Redi-Dri