• 主页
  • 查找结果
  • Use of a molybdenum(VI) complex as artificial protease in protein photocleavage.
Use of a molybdenum(VI) complex as artificial protease in protein photocleavage.
Journal of photochemistry and photobiology. B, Biology (2013-07-31)
Benchawan Jityuti, Teerayuth Liwporncharoenvong, Apinya Buranaprapuk
摘要
In this study, a molybdenum(VI) peroxo α-amino acid complex, MoO(O2)2(α-leucine) (H2O), was prepared and used as an artificial protease for site-specific cleavage of porcine pepsin, a model protein. Cleavage of pepsin by MoO(O2)2(α-leucine) (H2O) was achieved under photochemical conditions at room temperature and pH 7.0. The reaction was activated by irradiation of the MoO(O2)2(α-leucine) (H2O)-protein mixture by UV light (320 and 340nm) for up to 30min. No cleavage was observed in the absence of MoO(O2)2(α-leucine) (H2O) or the light. The photocleavage yield increased with irradiation time. The cleaved fragments were sequencable, and the cleavage site was assigned to Leu(112)-Tyr(113). The cleavage reaction was quenched by ethanol. Therefore, hydroxyl radicals may be involved in the reaction and responsible for the cleavage of the protein. This is the first demonstration of the successful photocleavage of proteins by a molybdenum complex. This observation can provide a new approach for the photochemical footprinting of metal binding sites on proteins.
材料
货号
品牌
产品描述
Sigma-Aldrich
胃蛋白酶 来源于猪胃粘膜, powder, ≥250 units/mg solid
Sigma-Aldrich
胃蛋白酶 来源于猪胃粘膜, lyophilized powder, ≥3,200 units/mg protein
Sigma-Aldrich
胃蛋白酶 来源于猪胃粘膜, lyophilized powder, ≥2,500 units/mg protein (E1%/280)
Sigma-Aldrich
胃蛋白酶 来源于猪胃粘膜, powder, ≥400 units/mg protein
Sigma-Aldrich
胃蛋白酶 来源于猪胃粘膜, powder, slightly beige, ≥500 U/mg
Sigma-Aldrich
胃蛋白酶 来源于猪胃粘膜, powder, slightly beige, 1200-2400 U/mg
Sigma-Aldrich
胃蛋白酶−琼脂糖 来源于猪胃粘膜, lyophilized powder, ≥30 units/mg dry solid
Sigma-Aldrich
胃蛋白酶 来源于猪胃粘膜, Suitable for manufacturing of diagnostic kits and reagents, lyophilized powder, ≥3200 units/mg protein