• 主页
  • 查找结果
  • Clinical and immunological outcome in cutaneous leishmaniasis patients treated with pentoxifylline.
Clinical and immunological outcome in cutaneous leishmaniasis patients treated with pentoxifylline.
The American journal of tropical medicine and hygiene (2014-02-26)
Graça Brito, Mayra Dourado, Ludmila Polari, Daniela Celestino, Lucas P Carvalho, Adriano Queiroz, Edgar M Carvalho, Paulo R L Machado, Sara Passos
摘要
Pentoxifylline is a tumor necrosis factor-α (TNF-α) inhibitor that also attenuates the immune response and decreases tissue inflammation. The association of pentoxifylline with antimony improves the cure rate of mucosal and cutaneous leishmaniasis. In this randomized and double blind pilot trial, cure rate was higher, although not significant, in patients who received antimony plus pentoxifylline than in those patients receiving antimony plus placebo. A significant decrease in TNF-α and interferon-γ (IFN-γ) levels during therapy was more pronounced in the antimony plus pentoxifylline group, whereas CCL-3 (Chemokine [C-C motif] ligand 3) decreased similarly in both groups. The increased levels of CXCL-9 (Chemokine [C-X-C motif] ligand 9) during therapy were lower in the antimony plus pentoxifylline group. Therapy with pentoxifylline modifies cytokines and chemokines production, which may be associated with therapeutic outcome.
材料
货号
品牌
产品描述
Sigma-Aldrich
锑, powder, -100 mesh, 99.5% trace metals basis
Sigma-Aldrich
锑, beads, 1-5 mm, low oxide, 99.999% trace metals basis
锑, rod, 20mm, diameter 4.7mm, 99.999%
锑, rod, 6mm, diameter 6mm, 99.999%
锑, foil, 50x50mm, thickness 2mm, hot-pressed
锑, foil, light tested, 25x25mm, thickness 0.05mm, permanent polyester support, 95+%
锑, foil, light tested, 50x50mm, thickness 0.05mm, permanent polyester support, 95+%
锑, foil, not light tested, 100x100mm, thickness 0.015mm, permanent polyester support, 95+%
锑, foil, not light tested, 100x100mm, thickness 0.01mm, permanent polyester support, 95+%
锑, foil, not light tested, 100x100mm, thickness 0.025mm, permanent polyester support, 95+%
锑, foil, not light tested, 100x100mm, thickness 0.05mm, permanent polyester support, 95+%
锑, foil, not light tested, 25x25mm, thickness 0.002mm, permanent polyester support, 95+%
锑, foil, not light tested, 25x25mm, thickness 0.005mm, permanent polyester support, 95+%
锑, foil, not light tested, 25x25mm, thickness 0.015mm, permanent polyester support, 95+%
锑, foil, not light tested, 25x25mm, thickness 0.025mm, permanent polyester support, 95+%
锑, foil, not light tested, 50x50mm, thickness 0.002mm, permanent polyester support, 95+%
锑, foil, not light tested, 50x50mm, thickness 0.005mm, permanent polyester support, 95+%
锑, foil, not light tested, 50x50mm, thickness 0.015mm, permanent polyester support, 95+%
锑, foil, not light tested, 50x50mm, thickness 0.025mm, permanent polyester support, 95+%
锑, foil, not light tested, 50x50mm, thickness 0.05mm, permanent polyester support, 95+%
锑, microfoil, 25x25mm, thinness 0.5μm, specific density 334.5μg/cm2, 6 micron aluminum permanent support, 95+%
锑, microfoil, 50x50mm, thinness 0.5μm, specific density 334.5μg/cm2, 6 micron aluminum permanent support, 95+%
锑, rod, 15mm, diameter 2.0mm, 99.99%
锑, rod, 25mm, diameter 2.0mm, 99.99%
锑, rod, 50mm, diameter 2.0mm, 99.99%
锑, rod, 50mm, diameter 4.7mm, 99.999%
锑, foil, not light tested, 25x25mm, thickness 0.01mm, permanent polyester support, 95+%
锑, foil, not light tested, 50x50mm, thickness 0.01mm, permanent polyester support, 95+%
社交媒体
LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon
Merck
科研、开发、生产。
作为生命科学行业的全球领先供应商,我们致力于为科研、生物技术开发和生产,以及制药药物疗法开发和生产提供各类解决方案和服务。
© 2021年版权归德国达姆施塔特默克集团(Merck KGaA)及/或其附属公司所有。版权所有。
未经许可,严禁复制本网站上的任何资料。