• 主页
  • 查找结果
  • ROI-scavenging enzyme activities as toxicity biomarkers in three species of marine microalgae exposed to model contaminants (copper, Irgarol and atrazine).
ROI-scavenging enzyme activities as toxicity biomarkers in three species of marine microalgae exposed to model contaminants (copper, Irgarol and atrazine).
Ecotoxicology and environmental safety (2014-04-15)
Pablo Lozano, Chiara Trombini, Elena Crespo, Julián Blasco, Ignacio Moreno-Garrido
摘要
There is a need to develop efficient tools to prevent damage to marine ecosystems due to pollution. Since microalgae play a key role in marine ecosystems, they are considered potentially useful for quick and sensitive toxicity bioassays. In this study an integrative analysis has been carried out of the anti-oxidant enzyme activities of marine microalgae species. Three marine microalgae species (Cylindrotheca closterium, a benthic diatom; Phaeodactylum tricornutum, a diatom which has been used as model organism in toxicity bioassays; and Rhodomonas salina, a cryptophyceae which is considered to present a certain level of heterotrophy) were exposed to selected concentrations of three model pollutants: copper (5 and 10µg L(-1)), atrazine (25 and 50µg L(-1)) and Irgarol (0.5 and 1.0µg L(-1)). These pollutant concentrations are environmentally relevant for coastal ecosystems, and have been selected for checking the efficiency of the reactive oxygen intermediate (ROI) scavenging enzyme system of these organisms. Superoxide dismutase (SOD), catalase (CAT), ascorbate peroxidase (APx) and glutathione peroxidase (GPx) activities were measured at the end of 24h exposure. The integrated biomarker response (IBR) index - in our case for oxidative stress - has been employed to evaluate the ROI-scavenging enzyme system for each species and each treatment. In general, the SOD and CAT enzyme activities measured were higher in exposed populations than in controls, whereas APx and GPx activities showed the opposite trend. These microalgae showed significant responses of oxidative stress biomarkers at environmentally relevant concentrations for the assayed pollutants and short exposure periods, conditions that most other model organisms cannot match. Therefore microalgae present clear advantages over other species for their prospective employment in an "early warning system".
材料
货号
品牌
产品描述
Sigma-Aldrich
羟丙基甲基纤维素, viscosity 2,600-5,600 cP, 2 % in H2O(20 °C)(lit.)
Sigma-Aldrich
铜, powder, <425 μm, 99.5% trace metals basis
Sigma-Aldrich
羟丙基甲基纤维素
Sigma-Aldrich
铜, foil, thickness 0.25 mm, 99.98% trace metals basis
Supelco
阿特拉津, PESTANAL®, analytical standard
Sigma-Aldrich
羟丙甲纤维素, meets USP testing specifications
Sigma-Aldrich
铜, powder, 99.999% trace metals basis
Sigma-Aldrich
羟丙基甲基纤维素, viscosity 80-120 cP, 2 % in H2O(20 °C)(lit.)
Sigma-Aldrich
羟丙基甲基纤维素, viscosity 40-60 cP, 2 % in H2O(20 °C)(lit.)
Sigma-Aldrich
铜, wire, diam. 1.0 mm, ≥99.9%
Sigma-Aldrich
羟丙基甲基纤维素, average Mn ~10,000
Sigma-Aldrich
铜, ACS reagent, granular, 10-40 mesh, ≥99.90%
Sigma-Aldrich
铜, nanopowder, 60-80 nm particle size (SAXS), ≥99.5% trace metals basis
Sigma-Aldrich
铜, powder, <75 μm, 99%
Sigma-Aldrich
铜, nanopowder, 40-60 nm particle size (SAXS), ≥99.5% trace metals basis
Supelco
阿特拉津, analytical standard
Sigma-Aldrich
羟丙基甲基纤维素, average Mn ~90,000
Sigma-Aldrich
铜, powder (spheroidal), 10-25 μm, 98%
Sigma-Aldrich
铜, foil, thickness 0.025 mm, 99.98% trace metals basis
Sigma-Aldrich
羟丙基甲基纤维素, average Mn ~86,000
Sigma-Aldrich
铜, foil, thickness 1.0 mm, 99.999% trace metals basis
Sigma-Aldrich
铜, foil, ≥99.8% (complexometric)
USP
羟丙基甲基纤维素, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
铜, powder (dendritic), <45 μm, 99.7% trace metals basis
Sigma-Aldrich
铜, foil, thickness 0.5 mm, 99.98% trace metals basis
Sigma-Aldrich
铜, turnings, purum p.a., ≥99.0%
Sigma-Aldrich
铜, bars (random sizes), ≥99.999% trace metals basis
Sigma-Aldrich
铜, rod, diam. 11 mm, 99.9999% trace metals basis
Supelco
阿特拉津, certified reference material, TraceCERT®
Sigma-Aldrich
铜, shot, −3-+14 mesh, 99%