Merck
  • 主页
  • 查找结果
  • Utilization of carbon nanotubes for the removal of rhodamine B dye from aqueous solutions.
Utilization of carbon nanotubes for the removal of rhodamine B dye from aqueous solutions.
Journal of nanoscience and nanotechnology (2014-04-18)
Sandeep Kumar, Gaurav Bhanjana, Kavita Jangra, Neeraj Dilbaghi, Ahmad Umar
摘要
Carbon nanotubes (CNTs) are attracting increasing research interest as promising adsorbents for harmful cations, anions, and other organic and inorganic impurities present in natural sources of water. This study examined the feasibility of removing Rhodamine B dye from aqueous solutions using multi walled carbon nanotubes (MWCNTs) synthesized by chemical vapor deposition (CVD) method. The effects of dye concentration, pH and contact time on adsorption of direct dye by CNTs were also evaluated. The study used the Langmuir and Temkin isotherms to describe equilibrium adsorption. Additionally, pseudo second-order model was adopted to evaluate experimental data and thereby elucidate the kinetic adsorption process. The adsorption percentage of dye increased as contact time increased. Conversely, the adsorption percentage of dye decreased as dye concentration increased. The pseudo second-order model best represented adsorption kinetics. The capacity of CNTs to adsorb Rhodamine B was 65-90% at different pH values.
材料
货号
品牌
产品描述
Sigma-Aldrich
水, Nuclease-Free Water, for Molecular Biology
Sigma-Aldrich
水, suitable for HPLC
Sigma-Aldrich
水, sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
水, HPLC Plus
Supelco
水, suitable for ion chromatography
Sigma-Aldrich
水, deionized
Sigma-Aldrich
水, for molecular biology, sterile filtered
Sigma-Aldrich
水, for embryo transfer, sterile-filtered, BioXtra, suitable for mouse embryo cell culture
Supelco
水, for TOC analysis
Sigma-Aldrich
罗丹明B, for fluorescence
Sigma-Aldrich
罗丹明B, ≥95% (HPLC)
Supelco
水, ACS reagent, for ultratrace analysis
纯水密度标准物质RM, 0.9982 g/mL(20 °C), UKAS ISO/IEC17025 and ISO Guide 34 certified, density: 0.9982 g/mL at 20 °C, density: 0.9970 g/mL at 25 °C
Sigma-Aldrich
水, BioPerformance Certified
Sigma-Aldrich
水, for cell biology, sterile ultrafiltered
Sigma-Aldrich
水, PCR Reagent
Sigma-Aldrich
水, ACS reagent
Supelco
罗丹明B, analytical standard
水, UKAS ISO/IEC17025 and ISO Guide 34 certified, density: 0.9982 g/mL at 20 °C, density: 0.9970 g/mL at 25 °C
Sigma-Aldrich
水, tested according to Ph. Eur.
Sigma-Aldrich
E-Toxate 水, endotoxin, free
Supelco
水, for HPCE, for luminescence, suitable for UV/Vis spectroscopy
Supelco
密度标准品 998kg/m3, H&D Fitzgerald Ltd. Quality
Sigma-Aldrich
低氘水, ≤1 ppm (Deuterium oxide)
Supelco
罗丹明 B 溶液, 0.2% in isopropanol, for TLC derivatization
Sigma-Aldrich
水-16O, ≥99.94 atom % 16O