  • Search Results
  • Gastric pepsin in middle ear fluid of children with otitis media: clinical implications.
Gastric pepsin in middle ear fluid of children with otitis media: clinical implications.
Current allergy and asthma reports (2008-10-23)
Zhaoping He, Robert C O'Reilly, Devendra Mehta
ABSTRACT
Gastroesophageal reflux and extraesophageal reflux have been postulated to be involved in the pathogenesis of otitis media. This is supported by recent studies revealing the presence of gastric pepsin in the middle ear space of children with otitis media but not in control patients without otitis media. Reflux's role in otitis media appears to be most pronounced in younger children and those with purulent effusions.
Sigma-Aldrich
胃蛋白酶 来源于猪胃粘膜, powder, ≥250 units/mg solid
Sigma-Aldrich
胃蛋白酶 来源于猪胃粘膜, lyophilized powder, ≥3,200 units/mg protein
Sigma-Aldrich
胃蛋白酶 来源于猪胃粘膜, powder, ≥400 units/mg protein
胃蛋白酶, powder, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
胃蛋白酶 来源于猪胃粘膜, powder, slightly beige, ≥500 U/mg
Sigma-Aldrich
胃蛋白酶 来源于猪胃粘膜, powder, slightly beige, 1200-2400 U/mg
Sigma-Aldrich
胃蛋白酶 来源于猪胃粘膜, Suitable for manufacturing of diagnostic kits and reagents, lyophilized powder, ≥3200 units/mg protein