• 主页
  • 查找结果
  • Factors influencing compact-extended structure equilibrium in oligomers of aβ1-40 peptide--an ion mobility mass spectrometry study.
Factors influencing compact-extended structure equilibrium in oligomers of aβ1-40 peptide--an ion mobility mass spectrometry study.
Journal of molecular biology (2014-05-27)
Ewa Sitkiewicz, Marcin Kłoniecki, Jarosław Poznański, Wojciech Bal, Michał Dadlez
摘要
Oligomers formed by amyloid β (Aβ) peptide are widely believed to be the main neurotoxic agent in Alzheimer's disease. Studies discovered a broad variety of oligomeric forms, which display different levels of toxicity. Some of these forms may further assemble into mature fibrils, while other might be off-pathway from conversion to fibrils and assemble into alternative forms. To better understand a relationship between the structure and toxicity of Aβ oligomers, we require systematic characterization and classification of all possible forms, facilitating rational design of the beneficial modifiers of their activity. In previous ion mobility analysis of Aβ1-40 oligomers, we have detected the coexistence of two alternative structural forms (compact and extended) in a pool of low-order Aβ1-40 oligomers. These forms may represent two pathways of the oligomer evolution, leading either to fibrils or to off-pathway oligomers, which are potential candidates for the neurotoxic species. Here, we have analyzed the impact of incubation time, the presence of selected metal ions and the effect of a series of point mutations on mutual population of alternative forms. We have shown that a salt bridge D23K28 provides stabilization of the compact form whereas G25 is required for the existence of the extended form. We have found that binding of metal ions also stabilizes the compact form. These results improve our understanding of the possible molecular mechanism of the bifurcation of structural evolution of non-monomeric Aβ species into an off-fibril pathway, ultimately leading to the formation of potentially neurotoxic species.
材料
货号
品牌
产品描述
Sigma-Aldrich
硫酸锌 七水合物, ACS reagent, 99%
Sigma-Aldrich
硫酸锌 七水合物, BioReagent, suitable for cell culture
Sigma-Aldrich
硫酸锌 七水合物, ReagentPlus®, ≥99.0%
Sigma-Aldrich
铜, powder, <425 μm, 99.5% trace metals basis
Sigma-Aldrich
铜, foil, thickness 0.25 mm, 99.98% trace metals basis
Sigma-Aldrich
硫酸锌 七水合物, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.5%
Sigma-Aldrich
铜, powder, 99.999% trace metals basis
Sigma-Aldrich
硫酸锌 溶液, BioUltra, for molecular biology, 2.0 M in H2O
Sigma-Aldrich
铜, wire, diam. 1.0 mm, ≥99.9%
Sigma-Aldrich
铜, ACS reagent, granular, 10-40 mesh, ≥99.90%
Sigma-Aldrich
铜, nanopowder, 60-80 nm particle size (SAXS), ≥99.5% trace metals basis
Sigma-Aldrich
硫酸锌 七水合物, ≥99.95% trace metals basis
Sigma-Aldrich
铜, powder, <75 μm, 99%
Sigma-Aldrich
铜, nanopowder, 40-60 nm particle size (SAXS), ≥99.5% trace metals basis
Sigma-Aldrich
铜, powder (spheroidal), 10-25 μm, 98%
Sigma-Aldrich
硫酸锌 溶液, 0.3 N
Sigma-Aldrich
硫酸锌 七水合物, meets USP testing specifications
Sigma-Aldrich
铜, foil, thickness 0.025 mm, 99.98% trace metals basis
Sigma-Aldrich
铜, foil, thickness 1.0 mm, 99.999% trace metals basis
Sigma-Aldrich
铜, foil, ≥99.8% (complexometric)
Sigma-Aldrich
铜, powder (dendritic), <45 μm, 99.7% trace metals basis
Sigma-Aldrich
铜, foil, thickness 0.5 mm, 99.98% trace metals basis
Sigma-Aldrich
铜, turnings, purum p.a., ≥99.0%
Sigma-Aldrich
铜, bars (random sizes), ≥99.999% trace metals basis
Sigma-Aldrich
铜, rod, diam. 11 mm, 99.9999% trace metals basis
Sigma-Aldrich
铜, shot, −3-+14 mesh, 99%
Sigma-Aldrich
铜, spherical shot, −20-+30 mesh, ≥99.5% trace metals basis
Sigma-Aldrich
铜, wire, diam. 0.25 mm, 99.999% trace metals basis
Sigma-Aldrich
铜, beads, 2-8 mm, ≥99.99% trace metals basis
Sigma-Aldrich
铜, electrolytic, Analytical Reagent, reag. Ph. Eur., ≥99.9% (complexometric), wire