• 主页
  • 查找结果
  • Effect of metals on Daphnia magna and cladocerans representatives of the Argentinean fluvial littoral.
Effect of metals on Daphnia magna and cladocerans representatives of the Argentinean fluvial littoral.
Journal of environmental biology (2014-07-10)
Regaldo Luciana, Ulises Reno, Susana Gervasio, Troiani Horacio, Ana María Gagneten
摘要
Chronic toxicity tests were conducted to assess the effect of Cu, Cr and Pb on Moinodaphnia macleayi and Ceriodaphnia dubia -two cladoceran species from the Argentinian Fluvial Littoral Zone (AFLZ)- and Daphnia magna -an holarctic species-. The specimens were exposed to three concentrations of each metal. As endpoints, the number of living and dead organisms, molts, neonates released, and the age of first reproduction were recorded. Chronic assays showed that Cu significantly affected the analyzed life history traits in the three species. The lowest Pb and Cr concentrations did not affect survival, molting or fecundity in D. magna. Conversely, in M. macleayi and C. dubia, survival, molting and fecundity showed highly significant differences in all the concentrations tested compared to control assay. The present study stresses the importance of using biological parameters as bioindicators, as well as the study species from the Southern Hemisphere to assess metal pollution.
材料
货号
品牌
产品描述
Sigma-Aldrich
铜, powder, <425 μm, 99.5% trace metals basis
Sigma-Aldrich
铜, foil, thickness 0.25 mm, 99.98% trace metals basis
Sigma-Aldrich
铬, chips, 99.995% trace metals basis
Sigma-Aldrich
铜, powder, 99.999% trace metals basis
Sigma-Aldrich
铜, wire, diam. 1.0 mm, ≥99.9%
Sigma-Aldrich
铜, ACS reagent, granular, 10-40 mesh, ≥99.90%
Sigma-Aldrich
铜, nanopowder, 60-80 nm particle size (SAXS), ≥99.5% trace metals basis
Sigma-Aldrich
铬, powder, ≥99% trace metals basis, <45 μm
Sigma-Aldrich
铜, powder (spheroidal), 10-25 μm, 98%
Sigma-Aldrich
铜, powder, <75 μm, 99%
Sigma-Aldrich
铜, foil, thickness 0.025 mm, 99.98% trace metals basis
Sigma-Aldrich
铜, foil, ≥99.8% (complexometric)
Sigma-Aldrich
铜, foil, thickness 1.0 mm, 99.999% trace metals basis
Sigma-Aldrich
铜, powder (dendritic), <45 μm, 99.7% trace metals basis
Sigma-Aldrich
铜, turnings, purum p.a., ≥99.0%
Sigma-Aldrich
铜, foil, thickness 0.5 mm, 99.98% trace metals basis
Sigma-Aldrich
铅, powder, −100 mesh, 99.95% trace metals basis
Sigma-Aldrich
铜, wire, diam. 0.25 mm, 99.999% trace metals basis
Sigma-Aldrich
铬, powder, 99.5%, −100 mesh
Sigma-Aldrich
铅, wire, diam. 1.0 mm, 99.99% trace metals basis
Sigma-Aldrich
铜, bars (random sizes), ≥99.999% trace metals basis
Sigma-Aldrich
铜, spherical shot, −20-+30 mesh, ≥99.5% trace metals basis
Sigma-Aldrich
铜, rod, diam. 11 mm, 99.9999% trace metals basis
Sigma-Aldrich
铜, shot, −3-+14 mesh, 99%
Sigma-Aldrich
铜, beads, 2-8 mm, ≥99.99% trace metals basis
Sigma-Aldrich
铅, powder, −325 mesh, ≥99% trace metals basis
Sigma-Aldrich
铜, electrolytic, Analytical Reagent, reag. Ph. Eur., ≥99.9% (complexometric), wire
Sigma-Aldrich
铜, beads, 2-8 mm, 99.9995% trace metals basis
Sigma-Aldrich
铜, wire, diam. 2.0 mm, 99.999% trace metals basis
Sigma-Aldrich
铅, shot, <2 mm, 99.9% trace metals basis