• 主页
  • 查找结果
  • Disentangling steric and electronic factors in monomeric bis(2-bromo-4-chloro-6-{[(2-hydroxyethyl)imino]methyl}phenolato-κ(2)N,O)copper(II).
Disentangling steric and electronic factors in monomeric bis(2-bromo-4-chloro-6-{[(2-hydroxyethyl)imino]methyl}phenolato-κ(2)N,O)copper(II).
Acta crystallographica. Section C, Structural chemistry (2014-07-06)
Piotr Zabierowski, Janusz Szklarzewicz, Wojciech Nitek
摘要
The title compound, [Cu(C9H8BrClNO2)2], is a square-planar complex. The potentially tridentate dibasic 2-bromo-4-chloro-6-{[(2-hydroxyethyl)imino]methyl}phenolate ligand coordinates in a trans-bis fashion to the Cu(II) centre via the imine N and phenolate O atoms. The Cu(II) atom lies on the centre of inversion of the molecule. The potentially coordinating hydroxyethyl group remains protonated and uncoordinated, taking part in intermolecular hydrogen bonds with vicinal groups, leading to the formation of a two-dimensional hydrogen-bond network with sheets parallel to the (101) plane. Substituent effects on the crystal packing and coordination modes of the ligand are discussed.
材料
货号
品牌
产品描述
Sigma-Aldrich
铜, powder, <425 μm, 99.5% trace metals basis
Sigma-Aldrich
铜, foil, thickness 0.25 mm, 99.98% trace metals basis
Sigma-Aldrich
铜, powder, 99.999% trace metals basis
Sigma-Aldrich
铜, wire, diam. 1.0 mm, ≥99.9%
Sigma-Aldrich
铜, ACS reagent, granular, 10-40 mesh, ≥99.90%
Sigma-Aldrich
铜, nanopowder, 60-80 nm particle size (SAXS), ≥99.5% trace metals basis
Sigma-Aldrich
铜, powder, <75 μm, 99%
Sigma-Aldrich
铜, nanopowder, 40-60 nm particle size (SAXS), ≥99.5% trace metals basis
Sigma-Aldrich
铜, powder (spheroidal), 10-25 μm, 98%
Sigma-Aldrich
铜, foil, thickness 0.025 mm, 99.98% trace metals basis
Sigma-Aldrich
铜, foil, thickness 1.0 mm, 99.999% trace metals basis
Sigma-Aldrich
铜, foil, ≥99.8% (complexometric)
Sigma-Aldrich
铜, powder (dendritic), <45 μm, 99.7% trace metals basis
Sigma-Aldrich
铜, foil, thickness 0.5 mm, 99.98% trace metals basis
Sigma-Aldrich
铜, turnings, purum p.a., ≥99.0%
Sigma-Aldrich
铜, bars (random sizes), ≥99.999% trace metals basis
Sigma-Aldrich
铜, rod, diam. 11 mm, 99.9999% trace metals basis
Sigma-Aldrich
铜, shot, −3-+14 mesh, 99%
Sigma-Aldrich
铜, spherical shot, −20-+30 mesh, ≥99.5% trace metals basis
Sigma-Aldrich
铜, wire, diam. 0.25 mm, 99.999% trace metals basis
Sigma-Aldrich
铜, beads, 2-8 mm, ≥99.99% trace metals basis
Sigma-Aldrich
铜, electrolytic, Analytical Reagent, reag. Ph. Eur., ≥99.9% (complexometric), wire
Sigma-Aldrich
铜, wire, diam. 2.0 mm, 99.999% trace metals basis
Sigma-Aldrich
铜, beads, 2-8 mm, 99.9995% trace metals basis
Sigma-Aldrich
铜, wire, diam. 0.64 mm, 99.995% trace metals basis
铜, foil, 150x150mm, thickness 0.1mm, annealed, 99.95+%
铜, rod, 200mm, diameter 4.8mm, as drawn, 99.99+%
铜, mesh, 150x150mm, nominal aperture 0.14mm, thickness 0.25mm, wire diameter 0.115mm, 100x100 wires/inch, open area 30.3%, plain weave mesh
铜, wire reel, 5m, diameter 0.05mm, as drawn, 99.99+%
铜, foil, 100x100mm, thickness 0.125mm, hard, 99.95+%