• 主页
  • 查找结果
  • Phytotoxicity and accumulation of chromium in carrot plants and the derivation of soil thresholds for Chinese soils.
Phytotoxicity and accumulation of chromium in carrot plants and the derivation of soil thresholds for Chinese soils.
Ecotoxicology and environmental safety (2014-07-30)
Changfeng Ding, Xiaogang Li, Taolin Zhang, Yibing Ma, Xingxiang Wang
摘要
Soil environmental quality standards in respect of heavy metals for farmlands should be established considering both their effects on crop yield and their accumulation in the edible part. A greenhouse experiment was conducted to investigate the effects of chromium (Cr) on biomass production and Cr accumulation in carrot plants grown in a wide range of soils. The results revealed that carrot yield significantly decreased in 18 of the total 20 soils with Cr addition being the soil environmental quality standard of China. The Cr content of carrot grown in the five soils with pH>8.0 exceeded the maximum allowable level (0.5mgkg(-1)) according to the Chinese General Standard for Contaminants in Foods. The relationship between carrot Cr concentration and soil pH could be well fitted (R(2)=0.70, P<0.0001) by a linear-linear segmented regression model. The addition of Cr to soil influenced carrot yield firstly rather than the food quality. The major soil factors controlling Cr phytotoxicity and the prediction models were further identified and developed using path analysis and stepwise multiple linear regression analysis. Soil Cr thresholds for phytotoxicity meanwhile ensuring food safety were then derived on the condition of 10 percent yield reduction.
材料
货号
品牌
产品描述
Sigma-Aldrich
铬, chips, 99.995% trace metals basis
Sigma-Aldrich
铬, powder, ≥99% trace metals basis, <45 μm
Sigma-Aldrich
铬, powder, 99.5%, −100 mesh
Sigma-Aldrich
铬, chips, thickness ~2 mm, 99.5%
铬, rod, 10mm, diameter 5.0mm, 99.7+%
铬, foil, light tested, 25x25mm, thickness 0.05mm, permanent polyester support, 99.99+%
铬, foil, light tested, 25x25mm, thickness 0.075mm, permanent polyester support, 99.99+%
铬, foil, not light tested, 100x100mm, thickness 0.0125mm, permanent polyester support, 99.99+%
铬, foil, not light tested, 100x100mm, thickness 0.015mm, permanent polyester support, 99.99+%
铬, foil, not light tested, 100x100mm, thickness 0.01mm, permanent polyester support, 99.99+%
铬, foil, not light tested, 100x100mm, thickness 0.025mm, permanent polyester support, 99.99+%
铬, foil, not light tested, 100x100mm, thickness 0.02mm, permanent polyester support, 99.99+%
铬, foil, not light tested, 100x100mm, thickness 0.03mm, permanent polyester support, 99.99+%
铬, foil, not light tested, 100x100mm, thickness 0.04mm, permanent polyester support, 99.99+%
铬, foil, not light tested, 25x25mm, thickness 0.0015mm, permanent polyester support, 99.99+%
铬, foil, not light tested, 25x25mm, thickness 0.001mm, permanent polyester support, 99.99+%
铬, foil, not light tested, 25x25mm, thickness 0.0025mm, permanent polyester support, 99.99+%
铬, foil, not light tested, 25x25mm, thickness 0.002mm, permanent polyester support, 99.99+%
铬, foil, not light tested, 25x25mm, thickness 0.003mm, permanent polyester support, 99.99+%
铬, foil, not light tested, 25x25mm, thickness 0.004mm, permanent polyester support, 99.99+%
铬, foil, not light tested, 25x25mm, thickness 0.005mm, permanent polyester support, 99.99+%
铬, foil, not light tested, 25x25mm, thickness 0.007mm, permanent polyester support, 99.99+%
铬, foil, not light tested, 25x25mm, thickness 0.009mm, permanent polyester support, 99.99+%
铬, foil, not light tested, 25x25mm, thickness 0.0125mm, permanent polyester support, 99.99+%
铬, foil, not light tested, 25x25mm, thickness 0.015mm, permanent polyester support, 99.99+%
铬, foil, not light tested, 25x25mm, thickness 0.01mm, permanent polyester support, 99.99+%
铬, foil, not light tested, 25x25mm, thickness 0.025mm, permanent polyester support, 99.99+%
铬, foil, not light tested, 25x25mm, thickness 0.03mm, permanent polyester support, 99.99+%
铬, foil, not light tested, 25x25mm, thickness 0.04mm, permanent polyester support, 99.99+%
铬, foil, not light tested, 25x25mm, thickness 0.05mm, permanent polyester support, 99.99+%