• 主页
  • 查找结果
  • Antioxidant response of Phragmites australis to Cu and Cd contamination.
Antioxidant response of Phragmites australis to Cu and Cd contamination.
Ecotoxicology and environmental safety (2014-09-07)
A Cristina S Rocha, C Marisa R Almeida, M Clara P Basto, M Teresa S D Vasconcelos
摘要
Metals are known to induce oxidative stress in plant cells. Antioxidant thiolic compounds are known to play an important role in plants׳ defence mechanisms against metal toxicity but, regarding salt marsh plants, their role is still very poorly understood. In this work, the involvement of non-protein thiols (NPT), such as cysteine (Cys), reduced glutathione (GSH), oxidised glutathione (GSSG) and total acid-soluble SH compounds (total thiols), in the tolerance mechanisms of the marsh plant Phragmites australis against Cu and Cd toxicity was assessed. Specimens of this plant, freshly harvested in an estuarine salt marsh, were exposed, for 7 days, to rhizosediment soaked with the respective elutriate contaminated with Cu (0, 10 and 100 mg/L) or Cd (0, 1, 10 mg/L). In terms of NPT production, Cu and Cd contamination induced different responses in P. australis. The content of Cys increased in plant tissue after plant exposure to Cu, whereas Cd contamination led to a decrease in GSSG levels. In general, metal contamination did not cause a significant variation on GSH levels. Both metals influenced, to some extent, the production of other thiolic compounds. Despite the accumulation of considerable amounts of Cu and Cd in belowground tissues, no visible toxicity signs were observed. So, antioxidant thiolic compounds were probably involved in the mechanisms used by P. australis to alleviate metal toxicity. As P. australis is considered suitable for phytostabilising metal-contaminated sediments, understanding its tolerance mechanisms to toxic metals is important to optimise the conditions for applying this plant in phytoremediation procedures.
材料
货号
品牌
产品描述
Sigma-Aldrich
L -还原型谷胱甘肽, ≥98.0%
Sigma-Aldrich
L-半胱氨酸, 97%
Sigma-Aldrich
L -氧化谷胱甘肽, ≥98% (HPLC)
Sigma-Aldrich
L-半胱氨酸, from non-animal source, BioReagent, suitable for cell culture, ≥98%
Sigma-Aldrich
L -还原型谷胱甘肽, BioReagent, suitable for cell culture, ≥98.0%, powder
Supelco
谷胱甘肽, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
L-半胱氨酸, BioUltra, ≥98.5% (RT)
Sigma-Aldrich
铜, powder, <425 μm, 99.5% trace metals basis
Sigma-Aldrich
L -还原型谷胱甘肽, BioXtra, ≥98.0%
Sigma-Aldrich
铜, foil, thickness 0.25 mm, 99.98% trace metals basis
Sigma-Aldrich
铜, powder, 99.999% trace metals basis
Sigma-Aldrich
L-半胱氨酸, ≥97%, FG
Sigma-Aldrich
镉, powder, −100 mesh, 99.5% trace metals basis
Sigma-Aldrich
铜, wire, diam. 1.0 mm, ≥99.9%
SAFC
L-半胱氨酸
Sigma-Aldrich
铜, ACS reagent, granular, 10-40 mesh, ≥99.90%
Sigma-Aldrich
铜, nanopowder, 60-80 nm particle size (SAXS), ≥99.5% trace metals basis
Sigma-Aldrich
L -氧化谷胱甘肽, ≥98%, lyophilized powder
Sigma-Aldrich
L -氧化谷胱甘肽, BioXtra, ≥98%
Sigma-Aldrich
铜, powder, <75 μm, 99%
Sigma-Aldrich
铜, nanopowder, 40-60 nm particle size (SAXS), ≥99.5% trace metals basis
Sigma-Aldrich
镉, granular, ≥99%, 5-20 mesh
谷胱甘肽, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
铜, powder (spheroidal), 10-25 μm, 98%
Supelco
L-半胱氨酸, certified reference material, TraceCERT®
Sigma-Aldrich
镉, granular, 30-80 mesh, ≥99%
Sigma-Aldrich
L-半胱氨酸, produced by Wacker Chemie AG, Burghausen, Germany, ≥98.0%
Sigma-Aldrich
铜, foil, thickness 0.025 mm, 99.98% trace metals basis
Sigma-Aldrich
铜, foil, thickness 1.0 mm, 99.999% trace metals basis
Sigma-Aldrich
铜, foil, ≥99.8% (complexometric)