• 主页
  • 查找结果
  • Tumor necrosis factor-like weak inducer of apoptosis or Fn14 deficiency reduce elastase perfusion-induced aortic abdominal aneurysm in mice.
Tumor necrosis factor-like weak inducer of apoptosis or Fn14 deficiency reduce elastase perfusion-induced aortic abdominal aneurysm in mice.
Journal of the American Heart Association (2014-08-06)
Carlos Tarín, Valvanera Fernández-Laso, Cristina Sastre, Julio Madrigal-Matute, Mónica Gómez, Carlos Zaragoza, Jesús Egido, Linda C Burkly, Jose L Martín-Ventura, Luis M Blanco-Colio
摘要
Abdominal aortic aneurysm (AAA) involves leukocyte recruitment, inflammatory cytokine production, vascular cell apoptosis, neovascularization, and vascular remodeling, all of which contribute to aortic dilatation. Tumor necrosis factor-like weak inducer of apoptosis (TWEAK) is a cytokine implicated in proinflammatory responses, angiogenesis, and matrix degradation but its role in AAA formation is currently unknown. Experimental AAA with aortic elastase perfusion in mice was induced in wild-type (WT), TWEAK deficient (TWEAK KO), or Fn14-deficient (Fn14 KO) mice. TWEAK or Fn14 KO deficiency reduced aortic expansion, lesion macrophages, CD3(+) T cells, neutrophils, CD31(+) microvessels, CCL2 and CCL5 chemokines expression, and MMP activity after 14 days postperfusion. TWEAK and Fn14 KO mice also showed a reduced loss of medial vascular smooth muscle cells (VSMC) that was related to a reduced number of apoptotic cells in these animals compared with WT mice. Aortas from WT animals present a higher disruption of the elastic layer and MMP activity than those from TWEAK or Fn14 KO mice, indicating a diminished vascular remodeling in KO animals. In vitro experiments unveiled that TWEAK induces CCL5 secretion and MMP-9 activation in both VSMC and bone marrow-derived macrophages, and decrease VSMC viability, effects dependent on Fn14. TWEAK/Fn14 axis participates in AAA formation by promoting lesion inflammatory cell accumulation, angiogenesis, matrix-degrading protease expression, and vascular remodeling. Blocking TWEAK/Fn14 interaction could be a new target for the treatment of AAA.
材料
货号
品牌
产品描述
Sigma-Aldrich
亚甲蓝, certified by the Biological Stain Commission
Sigma-Aldrich
L-赖氨酸 单盐酸盐, reagent grade, ≥98% (HPLC)
Sigma-Aldrich
亚甲蓝 溶液, for microscopy, concentrate according to Ehrlich, concentrated, aqueous solution
Sigma-Aldrich
弹性蛋白酶 来源于猪胰腺, Type I, ≥4.0 units/mg protein
Sigma-Aldrich
L-赖氨酸 单盐酸盐, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 98.5-101.0%
Sigma-Aldrich
ApopTag Peroxidase In Situ Apoptosis Detection Kit, The ApopTag Peroxidase In Situ Apoptosis Detection Kit detects apoptotic cells in situ by labeling & detecting DNA strand breaks by the TUNEL method.
Supelco
L-赖氨酸 单盐酸盐, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
亚甲蓝 溶液, 0.05 wt. % in H2O
Sigma-Aldrich
L-赖氨酸 单盐酸盐, BioUltra, ≥99.5% (AT)
赖氨酸 盐酸盐, European Pharmacopoeia (EP) Reference Standard
Supelco
L-赖氨酸盐酸盐 溶液, 100 mM amino acid in 0.1 M HCl, analytical standard
Supelco
L-赖氨酸 单盐酸盐, certified reference material, TraceCERT®
Sigma-Aldrich
亚甲蓝 溶液, 1.4 % (w/v) in 95% ethanol
Sigma-Aldrich
亚甲蓝 溶液, according to Loeffler