  • Search Results
  • Expression of a long variant of CRACR2A that belongs to the Rab GTPase protein family in endothelial cells.
Expression of a long variant of CRACR2A that belongs to the Rab GTPase protein family in endothelial cells.
Biochemical and biophysical research communications (2014-12-06)
Lesley A Wilson, Lynn McKeown, Sarka Tumova, Jing Li, David J Beech
ABSTRACT
CRACR2A protein is described in T cells as an EF-hand-containing modulator of calcium-release-activated calcium (CRAC) channels. Here we sought relevance to calcium entry of endothelial cells. Unexpectedly, short interfering RNA designed to deplete CRACR2A had no effect on CRAC channels in endothelial cells but reduced the abundance of a protein with about twice the mass of CRACR2A. Reference to gene sequence data indicated the potential for a variant transcript encoding a C-terminal Rab GTPase extension of CRACR2A. Full-length cloning demonstrated expression of the long variant in endothelial cells. It was designated CRACR2A-L. Sequence analysis suggested it to be a previously unrecognised member of the Rab GTPase family. It made a positive contribution to endothelial tube formation. The data suggest that endothelial cells contain a long variant of CRACR2A which is an EF-hand-containing Rab protein that lacks impact on CRAC channels.
Sigma-Aldrich
乙二胺四乙酸, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
乙二胺四乙酸 二钠盐 溶液, BioUltra, for molecular biology, pH 8.0, ~0.5 M in H2O
Sigma-Aldrich
苯甲磺酰氟, ≥98.5% (GC)
Sigma-Aldrich
乙二胺四乙酸, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
毒胡萝卜素, ≥98% (HPLC), solid film
Sigma-Aldrich
乙二胺四乙酸 溶液, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
乙二胺四乙酸, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
乙二胺四乙酸, purified grade, ≥98.5%, powder
Sigma-Aldrich
钙, granular, 99%
Sigma-Aldrich
苯甲磺酰氟, ≥99.0% (T)
Sigma-Aldrich
乙二胺四乙酸, 99.995% trace metals basis
Sigma-Aldrich
乙二胺四乙酸, BioUltra, ≥99.0% (KT)
Sigma-Aldrich
乙二胺四乙酸, ≥98.0% (KT)
Sigma-Aldrich
钙, dendritic pieces, purified by distillation, 99.99% trace metals basis
Sigma-Aldrich
腺苷 5'-三磷酸酶 来源于猪大脑皮质, lyophilized powder, ≥0.3 units/mg protein, pH 7.8
Sigma-Aldrich
钙, pieces, <1 cm, 99%
Sigma-Aldrich
钙, turnings, 99% trace metals basis
Sigma-Aldrich
钙, dendritic pieces, purified by distillation, 99.9% trace metals basis
Sigma-Aldrich
钙, dendritic pieces, purified by distillation, 99.5% trace metals basis