  • Search Results
  • Application of low tube voltage coronary CT angiography with low-dose iodine contrast agent in patients with a BMI of 26-30 kg/m2.
Application of low tube voltage coronary CT angiography with low-dose iodine contrast agent in patients with a BMI of 26-30 kg/m2.
Clinical radiology (2014-12-04)
G Sun, Y B Hou, B Zhang, L Yu, S X Li, L L Tan, D J Chen
ABSTRACT
To evaluate the feasibility of using a reduced radiation dose and reduced iodine intake (i.e., "double low": low tube voltage/low iodine dose contrast agent) scanning protocol and the adaptive iterative dose reduction (AIDR) reconstruction algorithm in coronary heart disease (CHD) patients with a BMI of 26-30 kg/m(2). One-hundred and seventy-nine consecutive CHD patients with a body mass index >26 kg/m² but <30 kg/m² were randomly assigned to two groups (group A: 53 men, 39 women, average age 61.83 ± 11.84 years, and group B: 40 men, 47 women, average age 62.25 ± 11.37 years) based on tube voltage, contrast agent, and algorithm used. Group A underwent the "double low" protocol (iodixanol at 270 mg iodine/ml, 100 kVp tube voltage, and AIDR). Group B received the conventional protocol [iopamidol at 370 mg iodine/ml, 120 kVp tube voltage, and filtered back projection (FBP)]. The volume CT dose index (CTDIvol), dose-length product (DLP), effective dose (ED), and iodine intake of patients in "double low" group A were significantly lower than the "conventional" group B (p < 0.001). The mean intraluminal attenuation and contrast enhancement in group A were also significantly less than group B (p < 0.001), whereas the image noise using AIDR in group A was significantly lower than group B using FBP (p < 0.001). However, the signal-to- noise ratio (SNR), contrast-to-noise ratio (CNR), and image-quality scores between the two groups were comparable. Use of 320-row CT with a "double low" scanning protocol for CCTA in patients with a BMI of 26-30 kg/m(2) not only provided images of diagnostic quality but also reduced both radiation dose and iodine intake during scanning.
Product Description
碘克沙醇, European Pharmacopoeia (EP) Reference Standard