  • Search Results
  • Functional analysis of porcine reproductive and respiratory syndrome virus N-glycans in infection of permissive cells.
Functional analysis of porcine reproductive and respiratory syndrome virus N-glycans in infection of permissive cells.
Virology (2015-02-11)
Juan Li, Michael P Murtaugh
ABSTRACT
The role of envelope protein-linked N-glycans in porcine reproductive and respiratory syndrome virus (PRRSV) infection of permissive cells was examined. N-acetylglucosamine (GlcNAc) and N-acetyllactosamine (LacNAc) oligomer-specific lectins bound to PRRSV and blocked virus attachment, resulting in reduced viral infection. However, addition of GlcNAc oligomers and LacNAc to cell culture together with PRRSV did not block infection. Removal or alteration of envelope protein-linked N-glycans also did not affect virus infection, indicating that PRRSV N-glycans are not required for virus infection. These findings show that steric hindrance of glycans on the PRRSV envelope by lectins or, presumably, other space-filling molecules, may interfere nonspecifically with infection by blocking protein interactions with cell surface receptors. Glycans themselves appear not to be required for infection of permissive cells, but may have important roles in avoidance of host immunity and in protein structure, intracellular virion growth and assembly.
Sigma-Aldrich
二甲基亚砜, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
二甲基亚砜, anhydrous, ≥99.9%
Sigma-Aldrich
二甲基亚砜, for molecular biology
Sigma-Aldrich
二甲基亚砜, ACS reagent, ≥99.9%
Sigma-Aldrich
羟乙基哌嗪乙硫磺酸, ≥99.5% (titration)
Sigma-Aldrich
二甲基亚砜, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
蔗糖, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
羟乙基哌嗪乙硫磺酸, BioPerformance Certified, ≥99.5% (titration), suitable for cell culture
Sigma-Aldrich
碳酸氢钠, powder, BioReagent, for molecular biology, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
碳酸氢钠, ACS reagent, ≥99.7%
Sigma-Aldrich
二甲基亚砜, suitable for HPLC, ≥99.7%
Sigma-Aldrich
二甲基亚砜, ReagentPlus®, ≥99.5%
Sigma-Aldrich
蔗糖, ≥99.5% (GC)
Sigma-Aldrich
二甲基亚砜, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
蔗糖, BioXtra, ≥99.5% (GC)
Sigma-Aldrich
乙二胺四乙酸, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
乙二胺四乙酸 二钠盐 溶液, BioUltra, for molecular biology, pH 8.0, ~0.5 M in H2O
Sigma-Aldrich
丙酮酸钠, ReagentPlus®, ≥99%
Sigma-Aldrich
蔗糖, BioUltra, for molecular biology, ≥99.5% (HPLC)
Sigma-Aldrich
磷酸钠, 96%
Supelco
蔗糖, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
蔗糖, puriss., meets analytical specification of Ph. Eur., BP, NF
Sigma-Aldrich
二甲基亚砜, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
二甲基亚砜, puriss. p.a., ACS reagent, ≥99.9% (GC)
Sigma-Aldrich
乙二胺四乙酸, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
丙酮酸钠, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99%
Sigma-Aldrich
HEPES缓冲溶液, 1 M in H2O
Sigma-Aldrich
碳酸氢钠, ReagentPlus®, ≥99.5%, powder
Sigma-Aldrich
乙二胺四乙酸 溶液, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
USP
蔗糖, United States Pharmacopeia (USP) Reference Standard