Merck
  • 主页
  • 查找结果
  • Synthesis of [¹⁸F]-labelled maltose derivatives as PET tracers for imaging bacterial infection.
Synthesis of [¹⁸F]-labelled maltose derivatives as PET tracers for imaging bacterial infection.
Molecular imaging and biology : MIB : the official publication of the Academy of Molecular Imaging (2014-10-04)
Mohammad Namavari, Gayatri Gowrishankar, Aileen Hoehne, Erwan Jouannot, Sanjiv S Gambhir
摘要
To develop novel positron emission tomography (PET) agents for visualization and therapy monitoring of bacterial infections. It is known that maltose and maltodextrins are energy sources for bacteria. Hence, (18)F-labelled maltose derivatives could be a valuable tool for imaging bacterial infections. We have developed methods to synthesize 4-O-(α-D-glucopyranosyl)-6-deoxy-6-[(18)F]fluoro-D-glucopyranoside (6-[(18)F]fluoromaltose) and 4-O-(α-D-glucopyranosyl)-1-deoxy-1-[(18)F]fluoro-D-glucopyranoside (1-[(18)F]fluoromaltose) as bacterial infection PET imaging agents. 6-[(18)F]fluoromaltose was prepared from precursor 1,2,3-tri-O-acetyl-4-O-(2',3',-di-O-acetyl-4',6'-benzylidene-α-D-glucopyranosyl)-6-deoxy-6-nosyl-D-glucopranoside (5). The synthesis involved the radio-fluorination of 5 followed by acidic and basic hydrolysis to give 6-[(18)F]fluoromaltose. In an analogous procedure, 1-[(18)F]fluoromaltose was synthesized from 2,3, 6-tri-O-acetyl-4-O-(2',3',4',6-tetra-O-acetyl-α-D-glucopyranosyl)-1-deoxy-1-O-triflyl-D-glucopranoside (9). Stability of 6-[(18)F]fluoromaltose in phosphate-buffered saline (PBS) and human and mouse serum at 37 °C was determined. Escherichia coli uptake of 6-[(18)F]fluoromaltose was examined. A reliable synthesis of 1- and 6-[(18)F]fluoromaltose has been accomplished with 4-6 and 5-8% radiochemical yields, respectively (decay-corrected with 95 % radiochemical purity). 6-[(18)F]fluoromaltose was sufficiently stable over the time span needed for PET studies (∼96% intact compound after 1-h and ∼65% after 2-h incubation in serum). Bacterial uptake experiments indicated that E. coli transports 6-[(18)F]fluoromaltose. Competition assays showed that the uptake of 6-[(18)F]fluoromaltose was completely blocked by co-incubation with 1 mM of the natural substrate maltose. We have successfully synthesized 1- and 6-[(18)F]fluoromaltose via direct fluorination of appropriate protected maltose precursors. Bacterial uptake experiments in E. coli and stability studies suggest a possible application of 6-[(18)F]fluoromaltose as a new PET imaging agent for visualization and monitoring of bacterial infections.
材料
货号
品牌
产品描述
Sigma-Aldrich
乙腈, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
水, Nuclease-Free Water, for Molecular Biology
Sigma-Aldrich
乙腈, anhydrous, 99.8%
Sigma-Aldrich
水, suitable for HPLC
Sigma-Aldrich
乙腈, HPLC Plus, ≥99.9%
Sigma-Aldrich
水, sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
水, HPLC Plus
Supelco
水, suitable for ion chromatography
Sigma-Aldrich
乙腈, ACS reagent, ≥99.5%
Sigma-Aldrich
水, deionized
Sigma-Aldrich
水, for molecular biology, sterile filtered
Sigma-Aldrich
乙腈, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
水, for embryo transfer, sterile-filtered, BioXtra, suitable for mouse embryo cell culture
Supelco
水, for TOC analysis
Sigma-Aldrich
乙腈 溶液, contains 0.1 % (v/v) trifluoroacetic acid, suitable for HPLC
Sigma-Aldrich
乙腈 溶液, contains 0.1 % (v/v) formic acid, suitable for HPLC
Supelco
水, ACS reagent, for ultratrace analysis
纯水密度标准物质RM, 0.9982 g/mL(20 °C), UKAS ISO/IEC17025 and ISO Guide 34 certified, density: 0.9982 g/mL at 20 °C, density: 0.9970 g/mL at 25 °C
Sigma-Aldrich
水, BioPerformance Certified
Sigma-Aldrich
水, for cell biology, sterile ultrafiltered
Sigma-Aldrich
乙腈, suitable for HPLC-GC, ≥99.8% (GC)
Sigma-Aldrich
乙腈, HPLC Plus, ≥99.9%, poly-coated bottles
Sigma-Aldrich
水, PCR Reagent
Sigma-Aldrich
乙腈, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99.5% (GC)
Supelco
乙腈, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
水, ACS reagent
Supelco
乙腈, analytical standard
水, UKAS ISO/IEC17025 and ISO Guide 34 certified, density: 0.9982 g/mL at 20 °C, density: 0.9970 g/mL at 25 °C
Sigma-Aldrich
乙腈, biotech. grade, ≥99.93%
Sigma-Aldrich
水, tested according to Ph. Eur.