• 主页
  • 查找结果
  • Silver nanoparticles alter proteoglycan expression in the promotion of tendon repair.
Silver nanoparticles alter proteoglycan expression in the promotion of tendon repair.
Nanomedicine : nanotechnology, biology, and medicine (2013-12-18)
Karen H L Kwan, Kelvin W K Yeung, Xuelai Liu, Kenneth K Y Wong, Ho Cheung Shum, Yun Wah Lam, Shuk Han Cheng, Kenneth M C Cheung, Michael K T To
摘要
This study demonstrates a novel method of using silver nanoparticles for Achilles tendon injury healing. In vitro results indicated a stimulatory effect on cell proliferation and collagen synthesis with silver nanoparticles. Biomechanical test on the 42-day post operation Achilles tendon sample exhibited a significant improvement in tensile modulus when compared to the untreated group. Histology suggested that silver nanoparticles promoted cell alignment and proteoglycan synthesis. The collagen deposition was also improved. An alleviation of tumor necrosis factor α, and an increase in fibromodulin and proliferating cell nuclear antigen expression were seen in silver nanoparticles group by immunohistochemistry. This study further corroborates the finding of our previous study that silver nanoparticles help to restore the functionality of injured connective tissues. We believe that the anti-inflammatory nature of silver nanoparticles has an important role in accelerating the healing process and reducing scarring, leading to better functional outcome. From the clinical editor: Tendon healing after surgeries remains a slow and tedious process, typically requiring several weeks of recovery time and gradual introduction of physical therapy. There are no currently utilized methods that could promote tendon healing. In this study, silver nanoparticles are reported to facilitate Achilles tendon repair in a model system, through increased proteoglycan and collagen synthesis, paving the way to potential clinical applications in the future.
材料
货号
品牌
产品描述
Sigma-Aldrich
甲醇, suitable for HPLC, ≥99.9%
Sigma-Aldrich
甲醇, anhydrous, 99.8%
Sigma-Aldrich
甲醇, HPLC Plus, ≥99.9%
Sigma-Aldrich
甲醇, HPLC Plus, ≥99.9%, poly-coated bottles
Sigma-Aldrich
甲醇, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
甲醇, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
甲醇, suitable for HPLC, gradient grade, 99.93%
Supelco
甲醇, analytical standard
Sigma-Aldrich
甲醇, ACS reagent, ≥99.8%
Sigma-Aldrich
氢氧化钠, reagent grade, ≥98%, pellets (anhydrous)
Sigma-Aldrich
氢氧化钠, ACS reagent, ≥97.0%, pellets
Sigma-Aldrich
氢氧化钠 溶液, 50% in H2O
Sigma-Aldrich
氢氧化钠, BioXtra, ≥98% (acidimetric), pellets (anhydrous)
Sigma-Aldrich
氢氧化钠 溶液, BioUltra, for molecular biology, 10 M in H2O
Sigma-Aldrich
氢氧化钠 溶液, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
甲醇, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)
USP
木精, United States Pharmacopeia (USP) Reference Standard
Supelco
氢氧化钠溶液, 0.1 M NaOH in water (0.1N), Eluent concentrate for IC
Sigma-Aldrich
氢氧化钠, puriss., meets analytical specification of Ph. Eur., BP, NF, E524, 98-100.5%, pellets
Supelco
氢氧化钠 溶液, 49-51% in water, eluent for IC
Sigma-Aldrich
甲醇, Laboratory Reagent, ≥99.6%
Supelco
甲醇, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
氢氧化钠, reagent grade, 97%, powder
Sigma-Aldrich
氢氧化钠 溶液, 5.0 M
Sigma-Aldrich
甲醇, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
氢氧化钠, puriss. p.a., ACS reagent, reag. Ph. Eur., K ≤0.02%, ≥98%, pellets
Sigma-Aldrich
甲醇, BioReagent, ≥99.93%
Sigma-Aldrich
氢氧化钠, pellets, semiconductor grade, 99.99% trace metals basis
Sigma-Aldrich
氢氧化钠, puriss. p.a., ACS reagent, K ≤0.02%, ≥98.0% (T), pellets
Sigma-Aldrich
甲醇, Absolute - Acetone free