• 主页
  • 查找结果
  • Higher insulin sensitivity and impaired insulin secretion in cachetic solid ehrlich tumour-bearing mice.
Higher insulin sensitivity and impaired insulin secretion in cachetic solid ehrlich tumour-bearing mice.
Hormone and metabolic research = Hormon- und Stoffwechselforschung = Hormones et metabolisme (2014-03-20)
N M Violato, A Rafacho, A C Boschero, J R Bosqueiro
摘要
Insulin secretion is mainly regulated by blood glucose concentration. On the other hand, changes in peripheral insulin sensitivity induce compensatory adaptations in pancreatic β-cells to maintain normoglycaemia. Tumour presence causes dramatic alterations in glucose homeostasis and insulin secretion because of the high glucose consumption by the tumour cells. Here, we investigated insulin secretion in solid Ehrlich tumour-bearing mice in association with cachexia. For that, male adult Swiss mice were subcutaneously inoculated with solid Ehrlich tumour cells and sacrificed at 14 days after tumour implantation (SET), while control mice received saline alone (CTL). Insulin secretion, following different stimuli, glucose tolerance, and insulin sensitivity as well as the expression of key proteins involved in insulin secretion was assessed. The SET group showed decreased glycaemia, insulinaemia, hepatic glycogen and body weight, and increased plasma free fatty acids and triglycerides, characteristics of cancer cachexia. A very interesting finding in this study was the development of higher glucose tolerance and insulin sensitivity in SET group. The dose-response curve of insulin secretion to increasing glucose concentrations (2.8-22.2 mM) showed an EC50 of 10 mM glucose for CTL mice and 13 mM glucose for SET mice. Insulin secretion was significantly reduced in SET islets at 30 mM KCl, 100 μM carbachol, 20 mM arginine, and 20 mM leucine. Moreover, AKT, PKA, PKC, and AchRM3 expressions were reduced by 17% to 24% in SET animals. These results, mainly the augmented insulin sensitivity, show that SET is an interesting model to study alterations in pancreatic function and carbohydrate metabolism in cancer cachexia.
材料
货号
品牌
产品描述
Sigma-Aldrich
羟乙基哌嗪乙硫磺酸, ≥99.5% (titration)
Sigma-Aldrich
碳酸氢钠, powder, BioReagent, for molecular biology, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
氯化钠, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
碳酸氢钠, ACS reagent, ≥99.7%
Sigma-Aldrich
羟乙基哌嗪乙硫磺酸, BioPerformance Certified, ≥99.5% (titration), suitable for cell culture
Sigma-Aldrich
氯化钠, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
氯化钠, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
氯化钠 溶液, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
氯化钠 溶液, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
HEPES缓冲溶液, 1 M in H2O
Sigma-Aldrich
氯化钠 溶液, 5 M
SAFC
氯化钠 溶液, 5 M
Sigma-Aldrich
碳酸氢钠, ReagentPlus®, ≥99.5%, powder
Sigma-Aldrich
碳酸氢钠, BioXtra, 99.5-100.5%
Sigma-Aldrich
氯化钠, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
SAFC
羟乙基哌嗪乙硫磺酸
Sigma-Aldrich
羟乙基哌嗪乙硫磺酸, BioUltra, for molecular biology, ≥99.5% (T)
Sigma-Aldrich
氯化钠, BioUltra, for molecular biology, ≥99.5% (AT)
Supelco
氯化钠, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
碳酸氢钠, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99.7%
Sigma-Aldrich
氯化钠 溶液, 0.85%
Sigma-Aldrich
氯化钠 溶液, BioUltra, for molecular biology, ~5 M in H2O
Sigma-Aldrich
氯化钠, 99.999% trace metals basis
Sigma-Aldrich
卡巴胆碱, ≥98% (titration), crystalline
Supelco
氯化钠, certified reference material for titrimetry, certified by BAM, ≥99.5%
SAFC
羟乙基哌嗪乙硫磺酸
Sigma-Aldrich
羟乙基哌嗪乙硫磺酸, BioXtra, pH 5.0-6.5 (1 M in H2O), ≥99.5% (titration)
Sigma-Aldrich
氯化钠, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Sigma-Aldrich
碳酸氢钠, puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, E500, 99.0-100.5%, powder
精氨酸, European Pharmacopoeia (EP) Reference Standard