  • Search Results
  • Short-term rosuvastatin treatment for the prevention of contrast-induced acute kidney injury in patients receiving moderate or high volumes of contrast media: a sub-analysis of the TRACK-D study.
Short-term rosuvastatin treatment for the prevention of contrast-induced acute kidney injury in patients receiving moderate or high volumes of contrast media: a sub-analysis of the TRACK-D study.
Chinese medical journal (2015-03-12)
Jian Zhang, Yi Li, Gui-Zhou Tao, Yun-Dai Chen, Tao-Hong Hu, Xue-Bin Cao, Quan-Min Jing, Xiao-Zeng Wang, Ying-Yan Ma, Geng Wang, Hai-Wei Liu, Bin Wang, Kai Xu, Jing Li, Jie Deng, Ya-Ling Han
ABSTRACT
Current randomized trials have demonstrated the effects of short-term rosuvastatin therapy in preventing contrast-induced acute kidney injury (CIAKI). However, the consistency of these effects on patients administered different volumes of contrast media is unknown. In the TRACK-D trial, 2998 patients with type 2 diabetes and concomitant chronic kidney disease (CKD) who underwent coronary/peripheral arterial angiography with or without percutaneous intervention were randomized to short-term (2 days before and 3 days after procedure) rosuvastatin therapy or standard-of-care. This prespecified analysis compared the effects of rosuvastatin versus standard therapy in patients exposed to (moderate contrast volume [MCV], 200-300 ml, n = 712) or (high contrast volume [HCV], ≥ 300 ml, n = 220). The primary outcome was the incidence of CIAKI. The secondary outcome was a composite of death, dialysis/hemofiltration or worsened heart failure at 30 days. Rosuvastatin treatment was associated with a significant reduction in CIAKI compared with the controls (2.1% vs. 4.4%, P = 0.050) in the overall cohort and in patients with MCV (1.7% vs. 4.5%, P = 0.029), whereas no benefit was observed in patients with HCV (3.4% vs. 3.9%, P = 0.834). The incidence of secondary outcomes was significantly lower in the rosuvastatin group compared with control group (2.7% vs. 5.3%, P = 0.049) in the overall cohort, but it was similar between the patients with MCV (2.0% vs. 4.2%, P = 0.081) or HCV (5.1% vs. 8.8%, P = 0.273). Periprocedural short-term rosuvastatin treatment is effective in reducing CIAKI and adverse clinical events for patients with diabetes and CKD after their exposure to a moderate volume of contrast medium.
Product Description
碘克沙醇, European Pharmacopoeia (EP) Reference Standard