  • Search Results
  • Bidirectional effects of methanol extract of Wei-Chang-An pill on gastrointestinal transit and the spasmolytic activity on isolated rat jejunum.
Bidirectional effects of methanol extract of Wei-Chang-An pill on gastrointestinal transit and the spasmolytic activity on isolated rat jejunum.
Journal of ethnopharmacology (2014-06-15)
Zhuo Qu, Jingze Zhang, Wenyuan Gao, Huimin Guo, Changxiao Liu
ABSTRACT
Wei-Chang-An pill (WCA pill), a traditional Chinese medicine, has been used for treating various gastrointestinal diseases for several decades. Despite the popular medicinal use of WCA pill, less data was available to its activity and mechanism in gastrointestinal disorders. To examine the effects of the methanol extract of WCA pill (ME) on gastrointestinal tract so as to assess some of the possible mechanisms involved in the clinical treatment. ME was studied on gastrointestinal transit in vivo including gastric emptying and small intestinal motility in normal and neostigmine-induced mice, as well as on the isolated tissue preparations of rat jejunum in vitro. In vivo, the gastric emptying decreased and intestinal transit increased after administration of ME in normal mice. However, administration of ME accelerated the intestinal transit ranging from 0.01 to 0.8 mg/mL and reduced it at the concentration of 1.6 and 3.2 mg/mL, while the gastric emptying was inhibited throughout the concentrations in neostigmine-induced mice. in vitro, ME caused inhibitory effect on the spontaneous contraction of rat-isolated jejunum in dose-dependent manner ranging from 0.01 to 6 mg/mL and also relaxed the acetylcholine chloride (Ach, 10(-6) M)-induced and K+ (60 mM)-induced contractions. ME shifted the Ca2+ concentration-response curves to right, similar to that caused by verapamil (0.025 mM). These results indicated that ME might play a bidirectional role in gastrointestinal transit modulation and the effects on isolated tissue are probably mediated through calcium influx and muscarinic receptors, which provides pharmacological basis for the clinical use of WCA pill in gastrointestinal tract disorders.
Sigma-Aldrich
乙腈, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
甲醇, suitable for HPLC, ≥99.9%
Sigma-Aldrich
乙腈, anhydrous, 99.8%
Sigma-Aldrich
甲醇, anhydrous, 99.8%
Sigma-Aldrich
乙腈, HPLC Plus, ≥99.9%
Sigma-Aldrich
甲醇, ACS reagent, ≥99.8%
Sigma-Aldrich
甲醇, HPLC Plus, ≥99.9%
Sigma-Aldrich
甲醇, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
乙腈, ACS reagent, ≥99.5%
Sigma-Aldrich
乙腈, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
甲醇, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)
Supelco
甲醇, analytical standard
Sigma-Aldrich
乙腈 溶液, contains 0.1 % (v/v) trifluoroacetic acid, suitable for HPLC
Sigma-Aldrich
乙腈 溶液, contains 0.1 % (v/v) formic acid, suitable for HPLC
Sigma-Aldrich
甲醇, Laboratory Reagent, ≥99.6%
Supelco
甲醇, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
甲醇, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
USP
木精, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
甲醇, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
甲醇, BioReagent, ≥99.93%
Sigma-Aldrich
甲醇, Absolute - Acetone free
Sigma-Aldrich
乙腈, suitable for HPLC-GC, ≥99.8% (GC)
Sigma-Aldrich
乙腈, HPLC Plus, ≥99.9%, poly-coated bottles
Sigma-Aldrich
甲醇, ACS reagent, ≥99.8%
Sigma-Aldrich
乙腈, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99.5% (GC)
Supelco
乙腈, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
甲醇, puriss., meets analytical specification of Ph Eur, ≥99.7% (GC)
Supelco
乙腈, analytical standard
Sigma-Aldrich
甲醇, HPLC Plus, ≥99.9%, poly-coated bottles
Sigma-Aldrich
乙腈, biotech. grade, ≥99.93%