  • Search Results
  • Dual tumor targeting with pH-sensitive and bioreducible polymer-complexed oncolytic adenovirus.
Dual tumor targeting with pH-sensitive and bioreducible polymer-complexed oncolytic adenovirus.
Biomaterials (2014-12-20)
Chang Yoon Moon, Joung-Woo Choi, Dayananda Kasala, Soo-Jung Jung, Sung Wan Kim, Chae-Ok Yun
ABSTRACT
Oncolytic adenoviruses (Ads) have shown great promise in cancer gene therapy but their efficacy has been compromised by potent immunological, biochemical, and specific tumor-targeting limitations. To take full advantage of the innate cancer-specific killing potency of oncolytic Ads but also exploit the subtleties of the tumor microenvironment, we have generated a pH-sensitive and bio-reducible polymer (PPCBA)-coated oncolytic Ad. Ad-PPCBA complexes showed higher cellular uptake at pH 6.0 than pH 7.4 in both high and low coxsackie and adenovirus receptor-(CAR)-expressing cells, thereby demonstrating Ad-PPCBA's ability to target the low pH hypoxic tumor microenvironment and overcome CAR dependence for target cell uptake. Endocytic mechanism studies indicated that Ad-PPCBA internalization is mediated by macropinocytosis instead of the CAR-dependent endocytic pathway that internalizes naked Ad. VEGF-specific shRNA-expressing oncolytic Ad complexed with PPCBA (RdB/shVEGF-PPCBA) elicited much more potent suppression of U87 human brain cancer cell VEGF gene expression in vitro, and human breast cancer MCF7 cell/Matrigel plug vascularization in a mouse model, when cancer cells had been previously infected at pH 6.0 versus pH 7.4. Moreover, intratumorally and intravenously injected RdB/shVEGF-PPCBA nanocomplexes elicited significantly higher therapeutic efficacy than naked virus in U87-tumor mouse xenograft models, reducing IL-6, ALT, and AST serum levels. These data demonstrated PPCBA's biocompatibility and capability to shield the Ad surface to prevent innate immune response against Ad after both intratumoral and systemic administration. Taken together, these results demonstrate that smart, tumor-specific, oncolytic Ad-PPCBA complexes can be exploited to treat both primary and metastatic tumors.
Sigma-Aldrich
二甲基亚砜, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
二甲基亚砜, for molecular biology
Sigma-Aldrich
二甲基亚砜, anhydrous, ≥99.9%
Sigma-Aldrich
二甲基亚砜, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
二甲基亚砜, ACS reagent, ≥99.9%
Sigma-Aldrich
二乙醚, contains 1 ppm BHT as inhibitor, anhydrous, ≥99.7%
Sigma-Aldrich
二甲基亚砜, suitable for HPLC, ≥99.7%
Sigma-Aldrich
二甲基亚砜, ReagentPlus®, ≥99.5%
Sigma-Aldrich
二甲基亚砜, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
二乙醚, anhydrous, ACS reagent, ≥99.0%, contains BHT as inhibitor
Sigma-Aldrich
噻唑蓝, 98%
Sigma-Aldrich
二乙醚, suitable for HPLC, ≥99.9%, inhibitor-free
Sigma-Aldrich
噻唑蓝, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥97.5% (HPLC)
Sigma-Aldrich
二乙醚, ACS reagent, anhydrous, ≥99.0%, contains BHT as inhibitor
Sigma-Aldrich
二甲基亚砜, puriss. p.a., ACS reagent, ≥99.9% (GC)
Sigma-Aldrich
二乙醚, contains BHT as inhibitor, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8%
Sigma-Aldrich
哌嗪, ReagentPlus®, 99%
Sigma-Aldrich
二甲基亚砜, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
二甲基亚砜, BioUltra, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
二甲基亚砜, PCR Reagent
Sigma-Aldrich
二甲基亚砜 溶液, 50 wt. % in H2O
Sigma-Aldrich
二乙醚, ACS reagent, ≥98.0%, contains ≤2% ethanol and ≤10ppm BHT as inhibitor
Supelco
二乙醚, analytical standard
Sigma-Aldrich
哌嗪, BioUltra, anhydrous, ≥99.0% (T)
Supelco
二甲基亚砜, for inorganic trace analysis, ≥99.99995% (metals basis)
Sigma-Aldrich
二乙醚, reagent grade, ≥98%, contains ≤2% ethanol and ≤10ppm BHT as inhibitor
Supelco
二甲基亚砜, analytical standard
USP
二甲基亚砜, United States Pharmacopeia (USP) Reference Standard
二甲基亚砜, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
二乙醚, puriss., contains ~5 mg/L 2,6-di-tert.-butyl-4-methylphenol as stabilizer, meets analytical specification of Ph. Eur., BP, ≥99.5% (GC)