Merck
Knockdown of EGFR inhibits growth and invasion of gastric cancer cells.
Cancer gene therapy (2014-11-15)
Y Zhen, L Guanghui, Z Xiefu
ABSTRACT
The epidermal growth factor receptor (EGFR) is an oncogenic trans-membranous receptor, which is overexpressed in multiple human cancers. However, the role of EGFR in gastric cancer (GC) is still elusive. In this study, we aimed to investigate the expression and molecular mechanisms of EGFR in GC cells. Forty cases of GC and the corresponding adjacent non-cancerous tissues (ANCT) were collected, and the expression of EGFR was assessed using immunohistochemistry in biopsy samples. Furthermore, EGFR signaling was blocked by constructed recombinant small hairpin RNA lentiviral vector (Lv-shRAGE) used to transfect into human GC SGC-7901 cells. The expression of AKT, proliferating cell nuclear antigen (PCNA) and matrix metallopeptidase-9 (MMP-9) was detected by real-time PCR and western blotting assays. Cell proliferative activities and invasive capability were, respectively, determined by MTT (3-[4,5-dimethylthiazol-2-yl]-2,5 diphenyl tetrazolium bromide) and Transwell assays. Cell apoptosis and cycle distribution were analyzed by flow cytometry. EGFR was found highly expressed in cancer tissues compared with the ANCT and correlated with lymph node metastases. Knockdown of EGFR reduced cell proliferation and invasion of GC with decreased expression of AKT, PCNA and MMP-9 and induced cell apoptosis and cycle arrest. Upregulation of EGFR expression is associated with lymph node metastases of GC, and blockade of EGFR signaling suppresses growth and invasion of GC cells through AKT pathway, suggesting that EGFR may represent a potential therapeutic target for this aggressive malignancy.
Sigma-Aldrich
牛血清白蛋白 来源于牛血清, lyophilized powder, ≥96% (agarose gel electrophoresis)
Sigma-Aldrich
过氧化氢 溶液, 30 % (w/w) in H2O, contains stabilizer
Sigma-Aldrich
过氧化氢 溶液, contains inhibitor, 30 wt. % in H2O, ACS reagent
Supelco
过氧化氢 溶液, ≥30%, for trace analysis
Millipore
过氧化氢 溶液, 3%, suitable for microbiology
Sigma-Aldrich
过氧化氢 溶液, 50 wt. % in H2O, stabilized
Supelco
过氧化氢 溶液, 30 % (w/w), for ultratrace analysis
Sigma-Aldrich
过氧化氢 溶液, contains ~200 ppm acetanilide as stabilizer, 3 wt. % in H2O
Sigma-Aldrich
过氧化氢 溶液, 34.5-36.5%
Sigma-Aldrich
过氧化氢溶液, 30% (w/w), puriss. p.a., reag. ISO, reag. Ph. Eur.
Sigma-Aldrich
过氧化氢 溶液, contains inhibitor, 30 wt. % in H2O, meets USP testing specifications
Sigma-Aldrich
DL-炔丙基甘氨酸, cystathionine γ-lyase inhibitor
Sigma-Aldrich
过氧化氢 溶液, purum p.a., ≥35% (RT)
Sigma-Aldrich
过氧化氢 溶液, contains inhibitor, 35 wt. % in H2O
Sigma-Aldrich
过氧化氢 溶液, contains potassium stannate as inhibitor, 30-32 wt. % in water, semiconductor grade, 99.999% trace metals basis
Sigma-Aldrich
过氧化氢 溶液, tested according to Ph. Eur.