  • Search Results
  • Study on the deformation of endothelial cells using a bio-inspired in vitro disease model.
Study on the deformation of endothelial cells using a bio-inspired in vitro disease model.
Microvascular research (2014-02-19)
Eunseok Seo, Kyung Won Seo, Ji-Won Park, Tae Geol Lee, Sang Joon Lee
ABSTRACT
A bio-inspired in vitro disease model was developed to investigate the basic biophysics of atherosclerotic diseases. In vivo study was conducted in advance using zebrafish fed with a normal diet and a cholesterol-enriched diet. The endothelial cells (ECs) of the zebrafishes fed with a normal diet are tightly attached and aligned. Their collagen has a fiber-like structure. By contrast, the endothelial cells of the zebrafish on high cholesterol diet are disorganized and their collagen has broken structures. In vitro models of human umbilical vein endothelial cells (HUVECs) were established on collagen films to mimic such in vivo experimental results. The normal collagen film simulates the extracellular matrix (ECM) in the blood vessels of a normal zebrafish, and the collagenase-treated collagen film mimics the ECM in blood vessels of an abnormal zebrafish. The HUVECs cultured on the normal collagen film are tightly attached, similar to those of a normal zebrafish. However, the cells cultured on the collagenase-treated collagen film are aggregated and biomarkers of endothelial dysfunction are expressed on the surface of the abnormal endothelial cells established on the denatured collagen film. The present in vitro model using a bio-inspired collagen film has a great potential for the design of novel therapies for clinical treatments of atherosclerosis through better understanding on the outbreak mechanism of atherosclerosis.
Sigma-Aldrich
二乙醚, contains 1 ppm BHT as inhibitor, anhydrous, ≥99.7%
Sigma-Aldrich
二乙醚, anhydrous, ACS reagent, ≥99.0%, contains BHT as inhibitor
Sigma-Aldrich
胆固醇, Sigma Grade, ≥99%
Sigma-Aldrich
二乙醚, suitable for HPLC, ≥99.9%, inhibitor-free
Sigma-Aldrich
二乙醚, ACS reagent, anhydrous, ≥99.0%, contains BHT as inhibitor
Sigma-Aldrich
二乙醚, contains BHT as inhibitor, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8%
Sigma-Aldrich
胆固醇, powder, BioReagent, suitable for cell culture, ≥99%
Sigma-Aldrich
氧氯化锆 八水合物, reagent grade, 98%
SAFC
胆固醇、植物的, SyntheChol®, NF, Ph. Eur., JP
Sigma-Aldrich
SyntheChol ® NS0 补充, 500 ×, synthetic cholesterol, animal component-free, aqueous solution, sterile-filtered, suitable for cell culture
Sigma-Aldrich
荧光素钠, for fluorescence, free acid
Sigma-Aldrich
胆固醇, from sheep wool, ≥92.5% (GC), powder
Supelco
胆固醇, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
二乙醚, ACS reagent, ≥98.0%, contains ≤2% ethanol and ≤10ppm BHT as inhibitor
Supelco
二乙醚, analytical standard
Supelco
胆固醇 溶液, certified reference material, 10 mg/mL in chloroform
Sigma-Aldrich
二乙醚, reagent grade, ≥98%, contains ≤2% ethanol and ≤10ppm BHT as inhibitor
Sigma-Aldrich
二乙醚, puriss., contains ~5 mg/L 2,6-di-tert.-butyl-4-methylphenol as stabilizer, meets analytical specification of Ph. Eur., BP, ≥99.5% (GC)
SAFC
胆固醇, from sheep wool, Controlled origin, meets USP/NF testing specifications
荧光素钠, European Pharmacopoeia (EP) Reference Standard