  • Search Results
  • Nanodisco balls: control over surface versus core loading of diagnostically active nanocrystals into polymer nanoparticles.
Nanodisco balls: control over surface versus core loading of diagnostically active nanocrystals into polymer nanoparticles.
ACS nano (2014-09-05)
Peter Chhour, Nicolas Gallo, Rabee Cheheltani, Dewight Williams, Ajlan Al-Zaki, Taejong Paik, Jessica L Nichol, Zhicheng Tian, Pratap C Naha, Walter R Witschey, Harry R Allcock, Christopher B Murray, Andrew Tsourkas, David P Cormode
ABSTRACT
Nanoparticles of complex architectures can have unique properties. Self-assembly of spherical nanocrystals is a high yielding route to such systems. In this study, we report the self-assembly of a polymer and nanocrystals into aggregates, where the location of the nanocrystals can be controlled to be either at the surface or in the core. These nanospheres, when surface decorated with nanocrystals, resemble disco balls, thus the term nanodisco balls. We studied the mechanism of this surface loading phenomenon and found it to be Ca(2+) dependent. We also investigated whether excess phospholipids could prevent nanocrystal adherence. We found surface loading to occur with a variety of nanocrystal types including iron oxide nanoparticles, quantum dots, and nanophosphors, as well as sizes (10-30 nm) and shapes. Additionally, surface loading occurred over a range of polymer molecular weights (∼30-3000 kDa) and phospholipid carbon tail length. We also show that nanocrystals remain diagnostically active after loading onto the polymer nanospheres, i.e., providing contrast in the case of magnetic resonance imaging for iron oxide nanoparticles and fluorescence for quantum dots. Last, we demonstrated that a fluorescently labeled protein model drug can be delivered by surface loaded nanospheres. We present a platform for contrast media delivery, with the unusual feature that the payload can be controllably localized to the core or the surface.
Sigma-Aldrich
四氢呋喃, anhydrous, ≥99.9%, inhibitor-free
Sigma-Aldrich
二氯甲烷, anhydrous, ≥99.8%, contains 40-150 ppm amylene as stabilizer
Sigma-Aldrich
三氟乙酸, ReagentPlus®, 99%
Sigma-Aldrich
三氟乙酸, suitable for HPLC, ≥99.0%
Sigma-Aldrich
四氢呋喃, anhydrous, contains 250 ppm BHT as inhibitor, ≥99.9%
Sigma-Aldrich
四氢呋喃, inhibitor-free, suitable for HPLC, ≥99.9%
Sigma-Aldrich
二氯甲烷, suitable for HPLC, ≥99.8%, contains amylene as stabilizer
Sigma-Aldrich
二乙醚, contains 1 ppm BHT as inhibitor, anhydrous, ≥99.7%
Sigma-Aldrich
二乙醚, anhydrous, ACS reagent, ≥99.0%, contains BHT as inhibitor
Sigma-Aldrich
二氯甲烷, contains 40-150 ppm amylene as stabilizer, ACS reagent, ≥99.5%
Sigma-Aldrich
二氯甲烷, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%, contains 50-150 ppm amylene as stabilizer
Sigma-Aldrich
二乙醚, suitable for HPLC, ≥99.9%, inhibitor-free
Sigma-Aldrich
四氢呋喃, contains 250 ppm BHT as inhibitor, ACS reagent, ≥99.0%
Sigma-Aldrich
氟化钠, ACS reagent, ≥99%
Sigma-Aldrich
二乙醚, ACS reagent, anhydrous, ≥99.0%, contains BHT as inhibitor
Sigma-Aldrich
三氟乙酸, puriss. p.a., suitable for HPLC, ≥99.0% (GC)
Sigma-Aldrich
二氯甲烷, puriss. p.a., ACS reagent, reag. ISO, ≥99.9% (GC)
Sigma-Aldrich
四氢呋喃, contains 250 ppm BHT as inhibitor, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99.9%
Sigma-Aldrich
二乙醚, contains BHT as inhibitor, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8%
Sigma-Aldrich
三氟乙酸, ≥99%, for protein sequencing
Sigma-Aldrich
二氯甲烷, suitable for HPLC, ≥99.9%, contains 40-150 ppm amylene as stabilizer
Sigma-Aldrich
四氢呋喃, suitable for HPLC, ≥99.9%, inhibitor-free
Sigma-Aldrich
二氯甲烷, ACS reagent, ≥99.5%, contains 40-150 ppm amylene as stabilizer
Supelco
二氯甲烷, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
氟化钠, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
二氯甲烷, analytical standard
Sigma-Aldrich
氟化钠, BioXtra, ≥99%
Sigma-Aldrich
四氢呋喃, ReagentPlus®, ≥99.0%, contains 250 ppm BHT as inhibitor
Supelco
三氟乙酸, analytical standard
Supelco
四氢呋喃, analytical standard