  • Search Results
  • Comparison of renal biomarkers with glomerular filtration rate in susceptibility to the detection of gentamicin-induced acute kidney injury in dogs.
Comparison of renal biomarkers with glomerular filtration rate in susceptibility to the detection of gentamicin-induced acute kidney injury in dogs.
Journal of comparative pathology (2014-08-05)
A Sasaki, Y Sasaki, R Iwama, S Shimamura, K Yabe, K Takasuna, T Ichijo, K Furuhama, H Satoh
ABSTRACT
Fourteen renal biomarkers were compared with measurement of glomerular filtration rate (GFR) in detecting acute kidney injury (AKI) in beagle dogs given gentamicin (40 mg/kg/day by subcutaneous injection) for 7 consecutive days. Serum and urinary biomarkers were measured before administration of gentamicin and then on days 4 and 8 after starting administration. GFR was derived by use of a simplified equation. Increased urinary cystatin C and decreased GFR occurred from day 4 and were detected before increases in blood urea nitrogen (BUN) and serum creatinine concentrations and changes in other urinary parameters. The closest correlation was between urinary cystatin C and GFR. At termination, microscopical examination revealed extensive necrosis of proximal tubular epithelium with hyaline casts in the kidney of treated dogs. These data indicate that urinary cystatin C is the most sensitive index of kidney injury and GFR reflects the kidney functional mass.
Sigma-Aldrich
碘, ACS reagent, ≥99.8%, solid
Sigma-Aldrich
碘, ≥99.99% trace metals basis
Sigma-Aldrich
碘, flakes, ReagentPlus®, ≥99%
Supelco
碘, ReagentPlus®, ≥99.8% (titration)
Sigma-Aldrich
碘, 99.999% trace metals basis
Sigma-Aldrich
碘, anhydrous, beads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
碘, puriss., meets analytical specification of Ph. Eur., BP, USP, 99.8-100.5%
碘克沙醇, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
碘, ReagentPlus®, 99.7% trace metals basis, beads, 1-3 mm
Sigma-Aldrich
碘, puriss., ≥99.5% (RT), particles (round)