  • Search Results
  • Structure-based mutational analysis of several sites in the E protein: implications for understanding the entry mechanism of Japanese encephalitis virus.
Structure-based mutational analysis of several sites in the E protein: implications for understanding the entry mechanism of Japanese encephalitis virus.
Journal of virology (2015-03-13)
Haibin Liu, Yi Liu, Shaobo Wang, Yanjun Zhang, Xiangyang Zu, Zheng Zhou, Bo Zhang, Gengfu Xiao
ABSTRACT
Japanese encephalitis virus (JEV), which causes viral encephalitis in humans, is a serious risk to global public health. The JEV envelope protein mediates the viral entry pathway, including receptor-binding and low-pH-triggered membrane fusion. Utilizing mutagenesis of a JEV infectious cDNA clone, mutations were introduced into the potential receptor-binding motif or into residues critical for membrane fusion in the envelope protein to systematically investigate the JEV entry mechanism. We conducted experiments evaluating infectious particle, recombinant viral particle, and virus-like particle production and found that most mutations impaired virus production. Subcellular fractionation confirmed that five mutations--in I0, ij, BC, and FG and the R9A substitution-impaired virus assembly, and the assembled virus particles of another five mutations--in kl and the E373A, F407A, L221S, and W217A substitutions--were not released into the secretory pathway. Next, we examined the entry activity of six mutations yielding infectious virus. The results showed N154 and the DE loop are not the only or major receptor-binding motifs for JEV entry into BHK-21 cells; four residues, H144, H319, T410, and Q258, participating in the domain I (DI)-DIII interaction or zippering reaction are important to maintain the efficiency of viral membrane fusion. By continuous passaging of mutants, adaptive mutations from negatively charged amino acids to positively charged or neutral amino acids, such as E138K and D389G, were selected and could restore the viral entry activity. Recently, there has been much interest in the entry mechanism of flaviviruses into host cells, including the viral entry pathway and membrane fusion mechanism. Our study provides strong evidence for the critical role of several residues in the envelope protein in the assembly, release, and entry of JEV, which also contributes to our understanding of the flaviviral entry mechanism. Furthermore, we demonstrate that the H144A, H319A, T410A, and Q258A mutants exhibit attenuated fusion competence, which may be used to develop novel vaccine candidates for flaviviruses.
Sigma-Aldrich
OptiPrep密度梯度培养基, used for cell and subcellular organelle isolation
Sigma-Aldrich
胆固醇, Sigma Grade, ≥99%
Sigma-Aldrich
苯甲磺酰氟, ≥98.5% (GC)
Sigma-Aldrich
胆固醇, powder, BioReagent, suitable for cell culture, ≥99%
SAFC
胆固醇、植物的, SyntheChol®, NF, Ph. Eur., JP
Sigma-Aldrich
SyntheChol ® NS0 补充, 500 ×, synthetic cholesterol, animal component-free, aqueous solution, sterile-filtered, suitable for cell culture
Sigma-Aldrich
苯甲磺酰氟, ≥99.0% (T)
Sigma-Aldrich
胆固醇, from sheep wool, ≥92.5% (GC), powder
Supelco
胆固醇, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
胆固醇 溶液, certified reference material, 10 mg/mL in chloroform
碘克沙醇, European Pharmacopoeia (EP) Reference Standard
SAFC
胆固醇, from sheep wool, Controlled origin, meets USP/NF testing specifications
Sigma-Aldrich
Anti-St. Louis Encephalitis Antibody, MSI-7, clone 6B6C-1, clone 6b6c-1, Chemicon®, from mouse