  • Search Results
  • Relating mRNA and protein biomarker levels in a Dehalococcoides and Methanospirillum-containing community.
Relating mRNA and protein biomarker levels in a Dehalococcoides and Methanospirillum-containing community.
Applied microbiology and biotechnology (2014-12-04)
Annette R Rowe, Cresten B Mansfeldt, Gretchen L Heavner, Ruth E Richardson
ABSTRACT
To better understand the quantitative relationships between messenger RNA (mRNA) and protein biomarkers relevant to bioremediation, we quantified and compared respiration-associated gene products in an anaerobic syntrophic community. Respiration biomarkers for Dehalococcoides, an organohalide reducer, and Methanospirillum, a hydrogenotrophic methanogen, were quantified via qRT-PCR for mRNA and multiple reaction monitoring (MRM) of proteotypic peptides for protein. mRNA transcripts of the Dehalococcoides reductive dehalogenases PceA, TceA, and DMC1545, and hydrogenase HupL, as well as the Methanospirillum oxidoreductases MvrD and FrcA were shown to be similarly regulated with respect to their temporal responses to substrate addition. However, MvrD was two orders of magnitude lower in mRNA abundance. Per cell, Dehalococcoides protein biomarkers quantified were more abundant than Methanospirillum proteins. Comparing mRNA with protein abundance, poor correlations were observed between mRNA transcript levels and the net protein produced. For example, Dehalococcoides HupL and TceA transcripts were similarly abundant though TceA was far more abundant at the protein level (167 ± 121 vs. 1095 ± 337 proteins per cell, respectively). In Methanospirillum, MvrD maintained comparable per-cell protein abundance to FrcA (42 ± 14 vs. 60 ± 1 proteins per cell, respectively) despite the significantly lower transcript levels. Though no variability in protein decay rates was observed, the mRNA translation rate quantified for TceA was greater than the other Dehalococcoides targets monitored. These data suggest that there is considerable variation in the relationship between mRNA abundance and protein production both across transcripts within an organism and across organisms. This highlights the importance of empirically based studies for interpreting biomarker levels in environmentally relevant organisms.
Sigma-Aldrich
乙腈, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
乙腈, anhydrous, 99.8%
Sigma-Aldrich
乙腈, HPLC Plus, ≥99.9%
Sigma-Aldrich
乙腈, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
乙腈, ACS reagent, ≥99.5%
Sigma-Aldrich
乙腈 溶液, contains 0.1 % (v/v) formic acid, suitable for HPLC
Sigma-Aldrich
乙腈 溶液, contains 0.1 % (v/v) trifluoroacetic acid, suitable for HPLC
Sigma-Aldrich
乙腈, HPLC Plus, ≥99.9%, poly-coated bottles
Sigma-Aldrich
乙腈, suitable for HPLC-GC, ≥99.8% (GC)
Supelco
乙腈, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
四氯乙烯, anhydrous, ≥99%
Sigma-Aldrich
碳,介孔, nanopowder, <500 nm particle size (DLS), >99.95% trace metals basis
Sigma-Aldrich
四氯乙烯, ACS reagent, ≥99.0%
Sigma-Aldrich
碳纳米纤维, graphitized (iron-free), composed of conical platelets, D × L 100 nm × 20-200 μm
Supelco
乙腈, analytical standard
Sigma-Aldrich
碳,介孔, average pore diameter 100±10 Å (typical), >99.95% trace metals basis
Sigma-Aldrich
活性炭 Norit®, Norit® SX ultra, from peat, corresponds U.S. Food chemicals codex (3rd Ed.), steam activated and acid washed, highly purified, powder
Sigma-Aldrich
碳纳米纤维, graphitized, platelets(conical), >98% carbon basis, D × L 100 nm × 20-200 μm
Sigma-Aldrich
乙腈, biotech. grade, ≥99.93%
Sigma-Aldrich
石墨化碳, nanopowder, graphitized, <500 nm particle size (DLS), >99.95% trace metals basis
Sigma-Aldrich
碳,介孔
Sigma-Aldrich
活性炭 Norit®, Norit® CA1, wood, chemically activated, powder
Sigma-Aldrich
活性炭 Norit®, Norit® GAC 1240W, from coal, for potable water processing, steam activated, granular
Sigma-Aldrich
碳纳米纤维, pyrolitically stripped, platelets(conical), >98% carbon basis, D × L 100 nm × 20-200 μm
Supelco
活性炭 Norit®, Norit® RBAA-3, rod
Supelco
四氯乙烯, analytical standard
Sigma-Aldrich
乙腈, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99.5% (GC)
Sigma-Aldrich
活性炭 Norit®, Norit® RB3, for gas purification, steam activated, rod
Sigma-Aldrich
甲烷, electronic grade, ≥99.998%
Sigma-Aldrich
乙腈, electronic grade, 99.999% trace metals basis