• 主页
  • 查找结果
  • Benzyl butyl phthalate induces migration, invasion, and angiogenesis of Huh7 hepatocellular carcinoma cells through nongenomic AhR/G-protein signaling.
Benzyl butyl phthalate induces migration, invasion, and angiogenesis of Huh7 hepatocellular carcinoma cells through nongenomic AhR/G-protein signaling.
BMC cancer (2014-08-02)
Cheng-Fang Tsai, Tsung-Hua Hsieh, Jau-Nan Lee, Chia-Yi Hsu, Yu-Chih Wang, Feng-Jie Lai, Kung-Kai Kuo, Hua-Lin Wu, Eing-Mei Tsai, Po-Lin Kuo
摘要
The widespread use of phthalates as plasticizers has raised public health concerns regarding their adverse effects, including an association with cancer. Although animal investigations have suggested an association between phthalate exposure and hepatocellular carcinoma, the mechanisms are unknown. The hepatocellular carcinoma cell line Huh7 was treated with benzyl butyl phthalate (BBP), and then analyzed by total internal reflection fluorescence microscopy, confocal microscopy and double immunogold transmission electron microscopy. Following BBP treatment, mRNA levels were measured by RT-PCR, protein levels were measured using western blot, and vascular endothelial growth factor levels were measured by an enzyme-linked immunosorbent assay. Cell migration and invasion assays were evaluated by transwell, and angiogenesis were performed by a tube formation assay. Nude mice were used to investigate metastasis and angiogenesis in vivo. BBP affected hepatocellular carcinoma progression through the aryl hydrocarbon receptor (AhR) and that benzyl butyl phthalate (BBP) stimulated AhR at the cell surface, which then interacted with G proteins and triggered a downstream signaling cascade. BBP activated AhR through a nongenomic action involving G-protein signaling rather than the classical genomic AhR action. BBP treatment promoted cell migration and invasion in vitro and metastasis in vivo via the AhR/Gβ/PI3K/Akt/NF-κB pathway. In addition, BBP induced both in vitro and in vivo angiogenesis through the AhR/ERK/VEGF pathway. These findings suggest a novel nongenomic AhR mechanism involving G-protein signaling induced by phthalates, which contributes to tumor progression of hepatocellular carcinoma.
材料
货号
品牌
产品描述
Sigma-Aldrich
羟乙基哌嗪乙硫磺酸, ≥99.5% (titration)
Sigma-Aldrich
D -(+)-葡萄糖, ≥99.5% (GC)
Sigma-Aldrich
DAPI, for nucleic acid staining
Sigma-Aldrich
羟乙基哌嗪乙硫磺酸, BioPerformance Certified, ≥99.5% (titration), suitable for cell culture
Sigma-Aldrich
D -(+)-葡萄糖, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.5%
Sigma-Aldrich
葡萄糖, meets EP, BP, JP, USP testing specifications, anhydrous
Sigma-Aldrich
氯化钙 溶液, BioUltra, for molecular biology, ~1 M in H2O
Sigma-Aldrich
D -(+)-葡萄糖, ACS reagent
Sigma-Aldrich
HEPES缓冲溶液, 1 M in H2O
Supelco
葡萄糖, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
D -(+)-葡萄糖, BioXtra, ≥99.5% (GC)
Sigma-Aldrich
氯化钙, meets USP testing specifications
Sigma-Aldrich
氯化钙, anhydrous, BioReagent, suitable for insect cell culture, suitable for plant cell culture, ≥96.0%
Sigma-Aldrich
氯化钙, anhydrous, powder, 99.99% trace metals basis
SAFC
羟乙基哌嗪乙硫磺酸
Sigma-Aldrich
羟乙基哌嗪乙硫磺酸, BioUltra, for molecular biology, ≥99.5% (T)
USP
右旋糖, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
D -(+)-葡萄糖, BioUltra, anhydrous, ≥99.5% (sum of enantiomers, HPLC)
Supelco
D -(+)-葡萄糖, analytical standard
SAFC
羟乙基哌嗪乙硫磺酸
Sigma-Aldrich
羟乙基哌嗪乙硫磺酸, BioXtra, pH 5.0-6.5 (1 M in H2O), ≥99.5% (titration)
Sigma-Aldrich
氯化钙, AnhydroBeads, −10 mesh, ≥99.9% trace metals basis
Sigma-Aldrich
D -(+)-葡萄糖, suitable for mouse embryo cell culture, ≥99.5% (GC)
Sigma-Aldrich
羟乙基哌嗪乙硫磺酸, BioXtra, suitable for mouse embryo cell culture, ≥99.5% (titration)
Supelco
用于 ISE 的钙离子溶液, 0.1 M Ca, analytical standard (for ion-selective electrodes)
Sigma-Aldrich
D -(+)-葡萄糖, tested according to Ph. Eur.
Sigma-Aldrich
羟乙基哌嗪乙硫磺酸, anhydrous, free-flowing, Redi-Dri, ≥99.5%
Sigma-Aldrich
D -(+)-葡萄糖, Hybri-Max, powder, BioReagent, suitable for hybridoma
Sigma-Aldrich
氯化钙, AnhydroBeads, −10 mesh, ≥99.99% trace metals basis
Supelco
HEPES, Pharmaceutical Secondary Standard; Certified Reference Material