  • Search Results
  • Lipid rafts couple class A scavenger receptors to phospholipase A2 activation during macrophage adhesion.
Lipid rafts couple class A scavenger receptors to phospholipase A2 activation during macrophage adhesion.
Journal of leukocyte biology (2014-07-30)
Shanthi Vadali, Steven R Post
ABSTRACT
SR-A mediated macrophage adhesion to modified ECM proteins in a process that involves physical attachment of SR-A to modified ECM and activation of Lyn-PI3K and PLA2-12/15-lipoxygenase signaling pathways. Structurally, SR-A-mediated cell adhesion requires a 6-aa membrane-proximal cytoplasmic motif. However, the mechanism that couples SR-A-mediated adhesion to activation of these distinct signaling pathways is not known. For other adhesion receptors, including integrins, localization in cholesterol-rich LRs is an important mechanism for coupling the receptor with the activation of specific signaling pathways. We hypothesized that SR-A-mediated macrophage adhesion might also involve LRs. Our results demonstrate that SR-A is enriched in LRs in HEK cells that heterologously express SR-A and in macrophages that endogenously expressed the receptor. We further show that a truncated SR-A construct (SR-A(Δ1-49)), which mediates cell adhesion but not ligand internalization, is also enriched in LRs, suggesting an association between LRs and SR-A-dependent cell adhesion. To examine this association more directly, we used the cholesterol chelator MβCD to deplete cholesterol and disrupt LR function. We found that cholesterol depletion significantly decreased SR-A-mediated macrophage adhesion. We further show that decreased SR-A-dependent macrophage adhesion following cholesterol depletion results from the inhibition of PLA2 but not PI3K activation. Overall, our results demonstrate an important role for LRs in selectively coupling SR-A with PLA2 activation during macrophage adhesion.
Sigma-Aldrich
胆固醇, Sigma Grade, ≥99%
Sigma-Aldrich
2-二(2-羟乙基)氨基-2-羟甲基-1,3-丙二醇, ≥98.0% (titration)
Sigma-Aldrich
胆固醇, powder, BioReagent, suitable for cell culture, ≥99%
Sigma-Aldrich
吲哚美辛, 98.5-100.5% (in accordance with EP)
SAFC
2-二(2-羟乙基)氨基-2-羟甲基-1,3-丙二醇
SAFC
胆固醇、植物的, SyntheChol®, NF, Ph. Eur., JP
Sigma-Aldrich
SyntheChol ® NS0 补充, 500 ×, synthetic cholesterol, animal component-free, aqueous solution, sterile-filtered, suitable for cell culture
Sigma-Aldrich
胆固醇, from sheep wool, ≥92.5% (GC), powder
Sigma-Aldrich
渥曼青霉素, from Penicillium funiculosum, ≥98% (HPLC and TLC)
Supelco
胆固醇, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
2-二(2-羟乙基)氨基-2-羟甲基-1,3-丙二醇, BioXtra, ≥98.0% (titration)
Supelco
吲哚美辛, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
2-二(2-羟乙基)氨基-2-羟甲基-1,3-丙二醇, BioUltra, ≥99.0% (NT)
Supelco
胆固醇 溶液, certified reference material, 10 mg/mL in chloroform
SAFC
2-二(2-羟乙基)氨基-2-羟甲基-1,3-丙二醇
Sigma-Aldrich
2-二(2-羟乙基)氨基-2-羟甲基-1,3-丙二醇, BioPerformance Certified, suitable for cell culture, suitable for insect cell culture, ≥98.0%
Sigma-Aldrich
吲哚美辛, meets USP testing specifications
碘克沙醇, European Pharmacopoeia (EP) Reference Standard
USP
吲哚美辛, United States Pharmacopeia (USP) Reference Standard
SAFC
胆固醇, from sheep wool, Controlled origin, meets USP/NF testing specifications
吲哚美辛, European Pharmacopoeia (EP) Reference Standard