  • Search Results
  • Development of a selective sorbent for the solid-phase extraction of terbuthylazine in olive oil samples: a molecular imprinting strategy.
Development of a selective sorbent for the solid-phase extraction of terbuthylazine in olive oil samples: a molecular imprinting strategy.
Journal of separation science (2015-02-07)
Raquel Garcia, Nuno Martins, Elisabete P Carreiro, Marco Simões, Maria M L Ribeiro Carrott, Peter J M Carrott, Anthony J Burke, Maria João Cabrita
ABSTRACT
Aiming to implement an analytical methodology that is highly selective for the extraction and quantification of terbuthylazine from olive oil, we successfully achieved: (i) the development of a molecularly imprinted polymer by bulk polymerization using terbuthylazine as template molecule, methacrylic acid as functional monomer, ethylene glycol dimethacrylate as cross-linker, and dichloromethane as porogen; (ii) characterization of the imprinting material using Fourier transform infrared spectroscopy, thermogravimetric analysis, nitrogen adsorption at 77 K, and scanning electron microscopy; (iii) their molecular recognition for the template molecule using high-performance liquid chromatography, and (iv) optimization of a solid-phase extraction procedure using as sorbent the synthesized molecularly imprinted polymer for the selective extraction and clean-up of terbuthylazine from spiked organic olive oil and further quantification of the pesticide levels by high-performance liquid chromatography. The suitability of the implemented analytical methodology was demonstrated, as concentrations of terbuthylazine below the tolerated maximum residue limits in the spiked organic olive oil samples could be satisfactorily analyzed with good precision/accuracy with high recovery rates (96%). Overall, the implemented methodology has proven to be reliable and robust and is highly promising in the field of sample preparation, particularly for the isolation/preconcentration of terbuthylazine in complex food samples.
Sigma-Aldrich
乙腈, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
甲醇, suitable for HPLC, ≥99.9%
Sigma-Aldrich
二氯甲烷, anhydrous, ≥99.8%, contains 40-150 ppm amylene as stabilizer
Sigma-Aldrich
乙腈, anhydrous, 99.8%
Sigma-Aldrich
甲醇, anhydrous, 99.8%
Sigma-Aldrich
二氯甲烷, suitable for HPLC, ≥99.8%, contains amylene as stabilizer
Sigma-Aldrich
乙酸, glacial, ReagentPlus®, ≥99%
Sigma-Aldrich
乙腈, HPLC Plus, ≥99.9%
Sigma-Aldrich
甲醇, ACS reagent, ≥99.8%
Sigma-Aldrich
乙酸, glacial, ACS reagent, ≥99.7%
Sigma-Aldrich
二氯甲烷, contains 40-150 ppm amylene as stabilizer, ACS reagent, ≥99.5%
Sigma-Aldrich
庚烷, suitable for HPLC, ≥99%
Sigma-Aldrich
甲醇, HPLC Plus, ≥99.9%
Sigma-Aldrich
甲醇, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
二氯甲烷, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%, contains 50-150 ppm amylene as stabilizer
Sigma-Aldrich
2,2′-偶氮(2-甲基丙腈), 98%
Sigma-Aldrich
庚烷, anhydrous, 99%
Sigma-Aldrich
庚烷, ReagentPlus®, 99%
Sigma-Aldrich
乙腈, ACS reagent, ≥99.5%
Sigma-Aldrich
乙酸, glacial, puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, 99.8-100.5%
Sigma-Aldrich
乙腈, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
二氯甲烷, puriss. p.a., ACS reagent, reag. ISO, ≥99.9% (GC)
Sigma-Aldrich
甲醇, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)
Sigma-Aldrich
乙二醇二甲基丙烯酸酯, 98%, contains 90-110 ppm monomethyl ether hydroquinone as inhibitor
Sigma-Aldrich
甲基丙烯酸, contains 250 ppm MEHQ as inhibitor, 99%
Sigma-Aldrich
乙酸, glacial, ≥99.99% trace metals basis
Supelco
甲醇, analytical standard
Sigma-Aldrich
乙酸 溶液, suitable for HPLC
Sigma-Aldrich
乙腈 溶液, contains 0.1 % (v/v) trifluoroacetic acid, suitable for HPLC
Sigma-Aldrich
乙腈 溶液, contains 0.1 % (v/v) formic acid, suitable for HPLC