Merck
  • 主页
  • 查找结果
  • Protection of cultured human hepatocytes from hydrogen peroxide‑induced apoptosis by relaxin‑3.
Protection of cultured human hepatocytes from hydrogen peroxide‑induced apoptosis by relaxin‑3.
Molecular medicine reports (2014-11-06)
Xiao Ma, Su Han, Wei Zhang, Yu-Jing Fan, Ming-Na Liu, Ai-Yun Liu, Bing-Rong Liu
摘要
Previous studies have suggested that hepatocyte apoptosis may be a fundamental underlying mechanism of liver injury and diseases, such as liver fibrosis. Relaxin‑3 has been reported to have anti‑fibrotic actions in the heart and to attenuate isoproterenol‑induced myocardial injury; however, the beneficial role of relaxin‑3 on hepatocyte apoptosis remains to be elucidated. The aim of the present study was to explore the role and possible mechanisms of relaxin‑3 through hydrogen peroxide (H2O2)‑induced apoptosis in primary human hepatocytes. Cells were treated with relaxin‑3 and then cell viability, morphological features, the presence of cleaved caspases as well as the levels of endoplasmic reticulum stress (ERS) protein markers and autophagy markers were evaluated. The H2O2 group showed significantly decreased cell viability, increased apoptosis as well as upregulation of caspases (cleaved caspase‑3, ‑8 and ‑9) and ERS protein markers compared with those of the control group. However, cells treated with relaxin‑3 (10 ng/ml) demonstrated improved cell viability, reduced apoptosis and decreased expression of cleaved caspases and ERS markers. However, the expression of autophagy markers remained unchanged following H2O2‑induced apoptosis and relaxin‑3 treatment. In conclusion, relaxin‑3 was shown to protect hepatocytes from H2O2‑induced apoptosis via downregulation of cleaved caspase‑8 and ‑9, as well as inhibition of the ERS pathway.
材料
货号
品牌
产品描述
Sigma-Aldrich
水, Nuclease-Free Water, for Molecular Biology
Sigma-Aldrich
水, suitable for HPLC
Sigma-Aldrich
水, sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
水, HPLC Plus
Sigma-Aldrich
噻唑蓝, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥97.5% (HPLC)
Sigma-Aldrich
噻唑蓝, 98%
Supelco
水, suitable for ion chromatography
Sigma-Aldrich
水, deionized
Sigma-Aldrich
水, for molecular biology, sterile filtered
Sigma-Aldrich
水, for embryo transfer, sterile-filtered, BioXtra, suitable for mouse embryo cell culture
Supelco
水, for TOC analysis
Supelco
水, ACS reagent, for ultratrace analysis
纯水密度标准物质RM, 0.9982 g/mL(20 °C), UKAS ISO/IEC17025 and ISO Guide 34 certified, density: 0.9982 g/mL at 20 °C, density: 0.9970 g/mL at 25 °C
Sigma-Aldrich
水, BioPerformance Certified
Sigma-Aldrich
水, for cell biology, sterile ultrafiltered
Sigma-Aldrich
水, PCR Reagent
Sigma-Aldrich
水, ACS reagent
水, UKAS ISO/IEC17025 and ISO Guide 34 certified, density: 0.9982 g/mL at 20 °C, density: 0.9970 g/mL at 25 °C
Sigma-Aldrich
水, tested according to Ph. Eur.
Sigma-Aldrich
E-Toxate 水, endotoxin, free
Supelco
水, for HPCE, for luminescence, suitable for UV/Vis spectroscopy
Supelco
密度标准品 998kg/m3, H&D Fitzgerald Ltd. Quality
Sigma-Aldrich
低氘水, ≤1 ppm (Deuterium oxide)
Sigma-Aldrich
水-16O, ≥99.94 atom % 16O