• 主页
  • 查找结果
  • Aberrant expression of claudin-4 and -7 in hepatocytes in the cirrhotic human liver.
Aberrant expression of claudin-4 and -7 in hepatocytes in the cirrhotic human liver.
Medical molecular morphology (2014-04-17)
Mitsuhiro Tsujiwaki, Masaki Murata, Akira Takasawa, Yutaro Hiratsuka, Rieko Fukuda, Kotaro Sugimoto, Yusuke Ono, Masanori Nojima, Satoshi Tanaka, Koichi Hirata, Takashi Kojima, Norimasa Sawada
摘要
The liver comprises hepatocytes and non-parenchymal cells such as bile duct epithelial cells. Claudin-4 and -7 are not expressed in hepatocytes under physiological conditions. It was reported that claudin-7 increased in human pulmonary fibroses. We therefore investigated claudin-4 and -7 expressions in human cirrhotic livers, in which hepatocyte proliferation is severely delayed. We examined liver tissues from 50 patients with liver tumors. The expression of claudin-4 and -7 in hepatocytes significantly increased with the grade of fibrosis, not with inflammatory activity, in the liver tissues of chronic hepatitis. The number of claudin-4- and -7-positive cells observed was greater than that of alpha-fetoprotein-positive hepatic progenitor cells. In primary cultures of mouse hepatocytes, the expression of claudin-4 and -7 was not induced by treatment with proinflammatory cytokines. In immunohistochemical analysis of liver tissues of 3,5-diethoxycarbonyl-1,4-dihydrocollidine-treated mice and primary cultures of mouse hepatocytes, the expression of claudin-4 and -7 increased with proliferation of progenitor cells. However, the claudin-4- and -7-positive cells were not always progenitor cells. Thus, claudin-4 and -7 were observed in hepatocytes of severely damaged mouse and human livers. These findings suggest that claudin-4- and -7-positive hepatocytes may exist during the process of differentiation from progenitor cells into mature hepatocytes.
材料
货号
品牌
产品描述
Sigma-Aldrich
二甲基亚砜, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
二甲基亚砜, for molecular biology
Sigma-Aldrich
二甲基亚砜, anhydrous, ≥99.9%
Sigma-Aldrich
乙酸, glacial, ReagentPlus®, ≥99%
Sigma-Aldrich
羟乙基哌嗪乙硫磺酸, ≥99.5% (titration)
Sigma-Aldrich
二甲基亚砜, ACS reagent, ≥99.9%
Sigma-Aldrich
二甲基亚砜, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
碳酸氢钠, powder, BioReagent, for molecular biology, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
乙酸, glacial, ACS reagent, ≥99.7%
Sigma-Aldrich
氯化钠, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
碳酸氢钠, ACS reagent, ≥99.7%
Sigma-Aldrich
羟乙基哌嗪乙硫磺酸, BioPerformance Certified, ≥99.5% (titration), suitable for cell culture
Sigma-Aldrich
地塞米松, powder, BioReagent, suitable for cell culture, ≥97%
Sigma-Aldrich
氯化钠, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
二甲基亚砜, suitable for HPLC, ≥99.7%
Sigma-Aldrich
二甲基亚砜, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
二甲基亚砜, ReagentPlus®, ≥99.5%
Sigma-Aldrich
氯化钠, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
氯化钠 溶液, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
氯化钠 溶液, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
乙二醇-双(2-氨基乙醚)-N,N,N′,N′-四乙酸, for molecular biology, ≥97.0%
Sigma-Aldrich
烟酰胺, BioReagent, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
乙酸, glacial, puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, 99.8-100.5%
Sigma-Aldrich
HEPES缓冲溶液, 1 M in H2O
Sigma-Aldrich
地塞米松, ≥98% (HPLC), powder
Sigma-Aldrich
氯化钠 溶液, 5 M
Sigma-Aldrich
L-脯氨酸, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
L-脯氨酸, from non-animal source, meets EP, USP testing specifications, suitable for cell culture
SAFC
氯化钠 溶液, 5 M
Sigma-Aldrich
碳酸氢钠, ReagentPlus®, ≥99.5%, powder