• 主页
  • 查找结果
  • Natural killer cells limit cardiac inflammation and fibrosis by halting eosinophil infiltration.
Natural killer cells limit cardiac inflammation and fibrosis by halting eosinophil infiltration.
The American journal of pathology (2015-01-28)
SuFey Ong, Davinna L Ligons, Jobert G Barin, Lei Wu, Monica V Talor, Nicola Diny, Jillian A Fontes, Elizabeth Gebremariam, David A Kass, Noel R Rose, Daniela Čiháková
摘要
Myocarditis is a leading cause of sudden cardiac failure in young adults. Natural killer (NK) cells, a subset of the innate lymphoid cell compartment, are protective in viral myocarditis. Herein, we demonstrated that these protective qualities extend to suppressing autoimmune inflammation. Experimental autoimmune myocarditis (EAM) was initiated in BALB/c mice by immunization with myocarditogenic peptide. During EAM, activated cardiac NK cells secreted interferon γ, perforin, and granzyme B, and expressed CD69, tumor necrosis factor-related apoptosis-inducing ligand treatment, and CD27 on their cell surfaces. The depletion of NK cells during EAM with anti-asialo GM1 antibody significantly increased myocarditis severity, and was accompanied by elevated fibrosis and a 10-fold increase in the percentage of cardiac-infiltrating eosinophils. The resultant influx of eosinophils to the heart was directly responsible for the increased disease severity in the absence of NK cells, because treatment with polyclonal antibody asialogangloside GM-1 did not augment myocarditis severity in eosinophil-deficient ΔdoubleGATA1 mice. We demonstrate that NK cells limit eosinophilic infiltration both indirectly, through altering eosinophil-related chemokine production by cardiac fibroblasts, and directly, by inducing eosinophil apoptosis in vitro. Altogether, we define a new pathway of eosinophilic regulation through interactions with NK cells.
材料
货号
品牌
产品描述
Sigma-Aldrich
羟乙基哌嗪乙硫磺酸, ≥99.5% (titration)
Sigma-Aldrich
羟乙基哌嗪乙硫磺酸, BioPerformance Certified, ≥99.5% (titration), suitable for cell culture
Sigma-Aldrich
乙二胺四乙酸 溶液, BioUltra, for molecular biology, pH 8.0, ~0.5 M in H2O
Sigma-Aldrich
乙二胺四乙酸, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
乙二胺四乙酸, anhydrous, BioUltra, ≥99% (titration)
Sigma-Aldrich
HEPES缓冲溶液, 1 M in H2O
Sigma-Aldrich
乙二胺四乙酸, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
乙二胺四乙酸 溶液, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
SAFC
羟乙基哌嗪乙硫磺酸
Sigma-Aldrich
乙二胺四乙酸, purified grade, ≥98.5%, powder
Sigma-Aldrich
羟乙基哌嗪乙硫磺酸, BioUltra, for molecular biology, ≥99.5% (T)
Sigma-Aldrich
乙二胺四乙酸, 99.995% trace metals basis
SAFC
羟乙基哌嗪乙硫磺酸
Sigma-Aldrich
羟乙基哌嗪乙硫磺酸, BioXtra, pH 5.0-6.5 (1 M in H2O), ≥99.5% (titration)
Sigma-Aldrich
偶氮二甲酸二乙酯 溶液, 40 wt. % in toluene
Sigma-Aldrich
乙二胺四乙酸, ≥98.0% (KT)
Sigma-Aldrich
羟乙基哌嗪乙硫磺酸, BioXtra, suitable for mouse embryo cell culture, ≥99.5% (titration)
Sigma-Aldrich
乙二胺四乙酸, BioUltra, ≥99.0% (KT)
Sigma-Aldrich
羟乙基哌嗪乙硫磺酸, anhydrous, free-flowing, Redi-Dri, ≥99.5%
Supelco
HEPES, Pharmaceutical Secondary Standard; Certified Reference Material