• 主页
  • 查找结果
  • Optical biosensor analysis in studying new synthesized bicalutamide analogs binding to androgen receptor.
Optical biosensor analysis in studying new synthesized bicalutamide analogs binding to androgen receptor.
Journal of pharmaceutical and biomedical analysis (2014-04-01)
Cecilia Fortugno, Greta Varchi, Andrea Guerrini, Pierre-Alain Carrupt, Carlo Bertucci
摘要
Bicalutamide (Casodex®) is a non-steroidal anti-androgen drug used in the treatment of prostate cancer, which represents the second most common malignancy diagnosed in men worldwide. In this work, we analyze the ability of some novel bicalutamide analogs to bind the androgen receptor, by using an optical biosensor. Androgen receptor was covalently immobilized on a carboxy methyl dextran matrix. The immobilized receptor chip was then used for the binding experiments of the bicalutamide analogs. The (R)-bicalutamide dissociation constant was in good agreement to the value reported in literature obtained by using radiolabeled targets. Most of the new synthesized compounds showed higher androgen receptor binding level, when compared to the reference. Our results clearly indicate that the surface plasmon resonance (SPR) technique offers many advantages with respect to other available technologies in terms of studying biomolecular interactions. Moreover, this study provides an effective methodology for determining the binding affinity of novel chemical entities for the isolated androgen receptor, thus excluding possible off-target interactions occurring in conventional cell-based techniques.
材料
货号
品牌
产品描述
Sigma-Aldrich
二甲基亚砜, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
二甲基亚砜, for molecular biology
Sigma-Aldrich
二甲基亚砜, anhydrous, ≥99.9%
Sigma-Aldrich
羟乙基哌嗪乙硫磺酸, ≥99.5% (titration)
Sigma-Aldrich
二甲基亚砜, ACS reagent, ≥99.9%
Sigma-Aldrich
二甲基亚砜, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
氯化钠, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
氢氧化钠, reagent grade, ≥98%, pellets (anhydrous)
Sigma-Aldrich
氯化钾, ACS reagent, 99.0-100.5%
Sigma-Aldrich
氢氧化钠, ACS reagent, ≥97.0%, pellets
Sigma-Aldrich
羟乙基哌嗪乙硫磺酸, BioPerformance Certified, ≥99.5% (titration), suitable for cell culture
Sigma-Aldrich
氢氧化钠 溶液, 50% in H2O
Sigma-Aldrich
乙酸钠, anhydrous, for molecular biology, ≥99%
Sigma-Aldrich
氢氧化钠, BioXtra, ≥98% (acidimetric), pellets (anhydrous)
Sigma-Aldrich
氯化钠, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
二甲基亚砜, suitable for HPLC, ≥99.7%
Sigma-Aldrich
氯化钠 溶液, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
二甲基亚砜, ReagentPlus®, ≥99.5%
Sigma-Aldrich
二甲基亚砜, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
氢氧化钠 溶液, BioUltra, for molecular biology, 10 M in H2O
Sigma-Aldrich
氯化钠, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
氢氧化钠 溶液, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
氯化钾, for molecular biology, ≥99.0%
Sigma-Aldrich
氯化钠 溶液, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
HEPES缓冲溶液, 1 M in H2O
Sigma-Aldrich
乙酸钠, anhydrous, ReagentPlus®, ≥99.0%
Sigma-Aldrich
氯化钠 溶液, 5 M
Supelco
氯化钾 溶液, for Ag/AgCl electrodes, ~3 M KCl, saturated with silver chloride
Sigma-Aldrich
氯化钾, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99.0%
Sigma-Aldrich
乙酸钠, ACS reagent, ≥99.0%