  • Search Results
  • Bioadhesive emulsions for control release of progesterone resistant to vaginal fluids clearance.
Bioadhesive emulsions for control release of progesterone resistant to vaginal fluids clearance.
International journal of pharmaceutics (2014-12-03)
Maria Campaña-Seoane, Aaron Peleteiro, Reyes Laguna, Francisco J Otero-Espinar
ABSTRACT
The aim of this study is to propose that mucoadhesive vaginal emulsions can be able to resist the clearance effect of vaginal fluid and to have an effective control release of progesterone. With this goal, silicon derivative, cyclomethicone pentamer, was selected as the bioadhesive and water resistant material. In order to obtain a system which is insensitive to the dilution of aqueous fluids, water in silicone (W/S) emulsions were prepared and different proportions of cyclomethicone as well as 8% or 15% w/w of progesterone were employed. The rheological, mechanical and mucoadhesive properties of emulsions were characterized and the drug release was measured for each formulation. Mucoadhesive behavior was determined and the influence of simulated vaginal fluid (SVF) at bioadhesion was assessed using three commercial mucoadhesive vaginal gels (Crinone(®), K-Y jelly(®) and Zidoval(®)) as the bioadhesive references. All assayed emulsions have good rheological and mechanical properties and their consistence and viscosity increase when the proportion of the internal phase increases. Related to mucoadhesion, in the absence of SVF, W/S emulsions showed similar bioadhesive levels like the commercial formulations. However, in the presence of SVF, W/S emulsions are able to keep their mucoadhesive properties while the marketed references drastically lose their consistency and adherence to the vaginal mucosa. Drug release profiles from W/S emulsion show that progesterone is released with pseudo-order zero kinetics and a constant release rate is maintained for at least two weeks. The results of the in vivo studies developed in rats show that after a single vaginal administration, bioadhesive W/S emulsions increase the uterine tissue progesterone levels in young and postmenopausal rats. Moreover in postmenopausal rats, they provide high uterine levels of progesterone compared to the bioadhesive-marketed gel used as a reference. Therefore, W/S emulsions have an interesting potential as bioadhesive vaginal delivery systems for drug administration.
Sigma-Aldrich
甘油, for molecular biology, ≥99.0%
Sigma-Aldrich
氯化钠, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
二乙醚, contains 1 ppm BHT as inhibitor, anhydrous, ≥99.7%
Sigma-Aldrich
甘油, ≥99.5%
Sigma-Aldrich
二乙醚, anhydrous, ACS reagent, ≥99.0%, contains BHT as inhibitor
Sigma-Aldrich
甘油, ACS reagent, ≥99.5%
Sigma-Aldrich
甘油, ReagentPlus®, ≥99.0% (GC)
Sigma-Aldrich
氯化钠, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
氯化钠 溶液, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
二乙醚, suitable for HPLC, ≥99.9%, inhibitor-free
Sigma-Aldrich
氯化钠 溶液, 0.9% in water, BioXtra, suitable for cell culture
SAFC
氯化钠 溶液, 5 M
Sigma-Aldrich
氯化钠, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
氯化钠 溶液, 5 M
Sigma-Aldrich
二乙醚, ACS reagent, anhydrous, ≥99.0%, contains BHT as inhibitor
Sigma-Aldrich
甘油, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for electrophoresis, ≥99% (GC)
Sigma-Aldrich
甘油, meets USP testing specifications
Sigma-Aldrich
甘油 溶液, 83.5-89.5% (T)
Supelco
甘油, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
氯化钠, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
二乙醚, contains BHT as inhibitor, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8%
Sigma-Aldrich
甘油, BioXtra, ≥99% (GC)
Sigma-Aldrich
氯化钠 溶液, BioUltra, for molecular biology, ~5 M in H2O
Sigma-Aldrich
氯化钠, BioUltra, for molecular biology, ≥99.5% (AT)
Sigma-Aldrich
黄体酮, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
黄体酮, ≥99%
Sigma-Aldrich
甘油, BioUltra, for molecular biology, anhydrous, ≥99.5% (GC)
Sigma-Aldrich
氯化钠 溶液, 0.85%
Supelco
氯化钠, Pharmaceutical Secondary Standard; Certified Reference Material
USP
甘油, United States Pharmacopeia (USP) Reference Standard