  • Search Results
  • NanoClusters surface area allows nanoparticle dissolution with microparticle properties.
NanoClusters surface area allows nanoparticle dissolution with microparticle properties.
Journal of pharmaceutical sciences (2014-05-03)
Christopher Kuehl, Nashwa El-Gendy, Cory Berkland
ABSTRACT
Poorly water-soluble drugs comprise the majority of new drug molecules. Nanoparticle agglomerates, called NanoClusters, can increase the dissolution rate of poorly soluble compounds by increasing particle surface area. Budesonide and danazol, two poorly soluble steroids, were studied as model compounds. NanoCluster suspensions were made using a Netzsch MiniCer media mill with samples collected between 5 and 15 h and lyophilized. Differential scanning calorimetry (DSC) and powder X-ray Diffraction were used to evaluate the physicochemical properties of the powders, and Brunauer, Emmett and Teller (BET) analysis was used to determine surface area. Scanning electron microscopy confirmed NanoClusters were between 1 and 5 μm. NanoCluster samples showed an increase in dissolution rate compared with the micronized stock and similar to a dried nanoparticle suspension. BET analysis determined an increase in surface area of eight times for budesonide NanoClusters and 10-15 times for danazol NanoClusters compared with the micronized stock. Melting temperatures decreased with increased mill time of NanoClusters by DSC. The increased surface area of NanoClusters provides a potential micron-sized alternative to nanoparticles to increase dissolution rate of poorly water-soluble drugs.
Sigma-Aldrich
乙腈, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
乙腈, anhydrous, 99.8%
Sigma-Aldrich
乙腈, HPLC Plus, ≥99.9%
Sigma-Aldrich
十二烷基硫酸钠, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC)
Sigma-Aldrich
十二烷基硫酸钠, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
十二烷基硫酸钠, ACS reagent, ≥99.0%
Sigma-Aldrich
十二烷基硫酸钠 溶液, BioUltra, for molecular biology, 10% in H2O
Sigma-Aldrich
乙腈, for HPLC, for UV, ≥99.9% (GC)
Sigma-Aldrich
十二烷基硫酸钠, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
乙腈, ACS reagent, ≥99.5%
Sigma-Aldrich
乙腈, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
十二烷基硫酸钠, BioUltra, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
十二烷基硫酸钠, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
十二烷基硫酸钠, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
十二烷基硫酸钠 溶液, BioUltra, for molecular biology, 20% in H2O
Sigma-Aldrich
乙腈 溶液, contains 0.1 % (v/v) trifluoroacetic acid, suitable for HPLC
Sigma-Aldrich
乙腈 溶液, contains 0.1 % (v/v) formic acid, suitable for HPLC
Supelco
十二烷基硫酸钠, suitable for ion pair chromatography, LiChropur, ≥99.0%
Supelco
十二烷基硫酸钠, dust-free pellets, suitable for electrophoresis, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
十二烷基硫酸钠, ≥90%
Sigma-Aldrich
乙腈, suitable for HPLC-GC, ≥99.8% (GC)
Sigma-Aldrich
乙腈, HPLC Plus, ≥99.9%, poly-coated bottles
Sigma-Aldrich
乙腈, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99.5% (GC)
Supelco
乙腈, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
十二烷基硫酸钠, ≥98.0% (GC)
Supelco
乙腈, analytical standard
十二烷基硫酸钠, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
十二烷基硫酸钠, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Sigma-Aldrich
乙腈, biotech. grade, ≥99.93%
Sigma-Aldrich
乙腈, electronic grade, 99.999% trace metals basis