• 主页
  • 查找结果
  • Cytosolic delivery via escape from the endosome using emulsion droplets and ultrasound.
Cytosolic delivery via escape from the endosome using emulsion droplets and ultrasound.
Journal of drug targeting (2015-02-13)
James R Lattin, Marjan Javadi, Michael McRae, William G Pitt
摘要
Vaporizing emulsion droplets may aid in endosomal rupture as a drug delivery route to the cytosol. Upon insonation, emulsion droplets formed from perfluorocarbon liquids may vaporize with sufficient expansion to disrupt liposomal or endosomal membranes. Emulsion droplets of perfluorohexane (PFC6) or perfluoropentane (PFC5) were prepared as free droplets in calcein or as droplets encapsulated within liposomes containing calcein. Folate-stimulated endocytosis created an experimental model, wherein calcein was self-quenched until released from the vesicles. Upon release, calcein was diluted below its self-quenching concentration and its release quantified by fluorescence. In this experimental model, folated emulsions or folated eLiposomes were incubated with folate-starved HeLa cells. Samples were exposed to two seconds of 20-kHz ultrasound (US) at 1 W/cm(2). Fluorescence microscopy identified released intracellular calcein. Upon insonation, both free emulsion samples and eLiposome samples produced calcein release to the cytosol. Calcein fluorescence was more intense in samples containing PFC5 compared to PFC6. Insonation of samples without emulsion droplets produced no cytosolic delivery. Likewise, cells that took up emulsion droplets but were not exposed to US did not exhibit fluorescence throughout the cell. These results suggest that vaporizing emulsion droplets are internalized into the cells and can produce endosomal escape of a therapeutic payload.
材料
货号
品牌
产品描述
Sigma-Aldrich
二甲基亚砜, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
纯乙醇, 200 proof, anhydrous, ≥99.5%
Sigma-Aldrich
纯乙醇, 190 proof, meets USP testing specifications
Sigma-Aldrich
纯乙醇, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
二甲基亚砜, for molecular biology
Sigma-Aldrich
乙醇, anhydrous, denatured
Sigma-Aldrich
二甲基亚砜, anhydrous, ≥99.9%
Sigma-Aldrich
二甲基亚砜, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
蔗糖, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
氯化钠, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
氯化钠, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
氯仿, anhydrous, ≥99%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
蔗糖, ≥99.5% (GC)
Sigma-Aldrich
二甲基亚砜, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
蔗糖, BioXtra, ≥99.5% (GC)
Sigma-Aldrich
氯化钠, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
氯化钠 溶液, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
N,N′-二环己基碳二亚胺, 99%
Sigma-Aldrich
氯化钠 溶液, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
氯仿, anhydrous, contains amylenes as stabilizer, ≥99%
Sigma-Aldrich
蔗糖, BioUltra, for molecular biology, ≥99.5% (HPLC)
Sigma-Aldrich
蔗糖, ≥99.5%
Sigma-Aldrich
叶酸, ≥97%
Sigma-Aldrich
氯化钠 溶液, 5 M
Sigma-Aldrich
蔗糖, puriss., meets analytical specification of Ph. Eur., BP, NF
SAFC
氯化钠 溶液, 5 M
Sigma-Aldrich
蔗糖, ACS reagent
Sigma-Aldrich
二甲基亚砜, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
叶酸, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥97%
Sigma-Aldrich
蔗糖, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99.5% (GC)