  • Search Results
  • Trauma/hemorrhagic shock instigates aberrant metabolic flux through glycolytic pathways, as revealed by preliminary (13)C-glucose labeling metabolomics.
Trauma/hemorrhagic shock instigates aberrant metabolic flux through glycolytic pathways, as revealed by preliminary (13)C-glucose labeling metabolomics.
Journal of translational medicine (2015-08-06)
Angelo D'Alessandro, Annie L Slaughter, Erik D Peltz, Ernest E Moore, Christopher C Silliman, Matthew Wither, Travis Nemkov, Anthony W Bacon, Miguel Fragoso, Anirban Banerjee, Kirk C Hansen
ABSTRACT
Metabolic derangement is a key hallmark of major traumatic injury. The recent introduction of mass spectrometry-based metabolomics technologies in the field of trauma shed new light on metabolic aberrations in plasma that are triggered by trauma and hemorrhagic shock. Alteration in metabolites associated with catabolism, acidosis and hyperglycemia have been identified. However, the mechanisms underlying fluxes driving such metabolic adaptations remain elusive. A bolus of U-(13)C-glucose was injected in Sprague-Dawley rats at different time points. Plasma extracts were analyzed via ultra-high performance liquid chromatography-mass spectrometry to detect quantitative fluctuations in metabolite levels as well as to trace the distribution of heavy labeled carbon isotopologues. Rats experiencing trauma did not show major plasma metabolic aberrations. However, trauma/hemorrhagic shock triggered severe metabolic derangement, resulting in increased glucose levels, lactate and carboxylic acid accumulation. Isotopologue distributions in late Krebs cycle metabolites (especially succinate) suggested a blockade at complex I and II of the electron transport chain, likely due to mitochondrial uncoupling. Urate increased after trauma and hemorrhage. Increased levels of unlabeled mannitol and citramalate, metabolites of potential bacterial origin, were also observed in trauma/hemorrhagic shock rats, but not trauma alone or controls. These preliminary results are consistent with observations we have recently obtained in humans, and expand upon our early results on rodent models of trauma and hemorrhagic shock by providing the kinetics of glucose fluxes after trauma and hemorrhage. Despite the preliminary nature of this study, owing to the limited number of biological replicates, results highlight a role for shock, rather than trauma alone, in eliciting systemic metabolic aberrations. This study provides the foundation for tracing experiments in rat models of trauma. The goal is to improve our understanding of substrate specific metabolic derangements in trauma/hemorrhagic shock, so as to design resuscitative strategies tailored toward metabolic alterations and the severity of trauma.
Sigma-Aldrich
水, Nuclease-Free Water, for Molecular Biology
Sigma-Aldrich
乙腈, anhydrous, 99.8%
Sigma-Aldrich
水, sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
水, for molecular biology, sterile filtered
Sigma-Aldrich
水, for embryo transfer, sterile-filtered, BioXtra, suitable for mouse embryo cell culture
Sigma-Aldrich
乙腈 溶液, contains 0.1 % (v/v) trifluoroacetic acid, suitable for HPLC
Sigma-Aldrich
乙腈 溶液, contains 0.1 % (v/v) formic acid, suitable for HPLC
Sigma-Aldrich
水, BioPerformance Certified
Sigma-Aldrich
水, for cell biology, sterile ultrafiltered
Sigma-Aldrich
乙腈, HPLC Plus, ≥99.9%, poly-coated bottles
Sigma-Aldrich
水, PCR Reagent
Sigma-Aldrich
E-Toxate 水, endotoxin, free
Sigma-Aldrich
乙腈, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
低氘水, ≤1 ppm (Deuterium oxide)
Sigma-Aldrich
乙腈, ≥99.5%, ACS reagent
Sigma-Aldrich
乙腈 溶液, contains 0.05 % (v/v) trifluoroacetic acid
Sigma-Aldrich
乙腈 溶液, contains 10.0% acetone, 40.0% 2-propanol, 0.05% formic acid
Sigma-Aldrich
乙腈
Sigma-Aldrich
乙腈 溶液, contains 0.05 % (w/v) ammonium formate, 0.1 % (v/v) formic acid, 5 % (v/v) water, suitable for HPLC
D-Glucose-13C6,1,2,3,4,5,6,6-d7, 77 atom % D, 99 atom % 13C, 99% (CP)
Sigma-Aldrich
D-葡萄糖-3-13C, 99 atom % 13C, 99% (CP)
Supelco
乙腈, HPLC grade, ≥99.93%
Sigma-Aldrich
水-16O, ≥99.94 atom % 16O
Sigma-Aldrich
D-Glucose-13C6,1,2,3,4,5,6,6-d7, 99 atom % 13C, 97 atom % D
Sigma-Aldrich
乙腈