  • Search Results
  • Development and validation of an improved HPLC-MS/MS method for comparative pharmacokinetics of penehyclidine hydrochloride following a single intravenous or intramuscular injection.
Development and validation of an improved HPLC-MS/MS method for comparative pharmacokinetics of penehyclidine hydrochloride following a single intravenous or intramuscular injection.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences (2015-10-08)
Libo Zhao, Yannan Zang, Yuwen Sun, Zhongna Wei, Shan Jing, Qian Wang, Youzhong An, Yi Fang, Wanyu Feng
ABSTRACT
Penehyclidine hydrochloride (PHC) is an anticholinergic drug with both antimuscarinic and antinicotinic activity. In order to compare the pharmacokinetics of two administration routes (intravenous injection (i.v.) and intramuscular injection (i.m.)) of PHC, an improved High Performance Liquid Chromatography Tandem Mass Spectrometry (HPLC-MS/MS) bioanalytical method was developed for the quantification of PHC in plasma and urine using verapamil as the internal standard (I.S.). Chromatography was performed using a Thermo Hypersil GOLD column (30mm×2.1mm, 3μm), with a gradient elution of 1‰ formic acid-10mmol/L ammonium acetate and acetonitrile at 0.3mL/min. Detection and quantitation were performed by electrospray ionization (ESI) and multiple reaction monitoring (MRM) in the positive ion mode. The most intense [M+H](+) MRM transition of PHC at m/z 316.2→128.3 was used for PHC quantitation, and the transition at m/z 454.6→303.2 was used to monitor I.S. The lower limit of quantification (LLOQ) was 0.05 ng/mL. The intraday precision was <6.71% and the interday precision was <11.69%. The pharmacokinetic parameters of i.v. and i.m. administration routes were as follows (i.v. vs i.m.): t1/2 15.73 vs 17.24h, Tmax 0.06 vs 0.26h, AUC0-t 69.35 vs 67.90hng/mL, AUC0-inf 78.24 vs 79.67hng/mL, Cmax 37.5 vs 9.1ng/mL, Ae0-24h 22.7 vs 25.21μg. There were no significant differences between parameters t1/2 and AUC (P>0.05), but significant differences were observed in Cmax, Tmax and Ae0-24h between the two administration routes (P<0.05). The mean absolute bioavailability of the i.m. administration route was 98.4% (95% confidence interval, 93.4-103.6%). Safety results showed that PHC appeared to be well tolerated in both i.v. and i.m. administration routes and pharmacokinetic results showed that PHC was nearly completely absorbed via i.m. administration route.
Sigma-Aldrich
乙腈, anhydrous, 99.8%
Sigma-Aldrich
甲醇, anhydrous, 99.8%
Sigma-Aldrich
甲醇, ACS reagent, ≥99.8%
Sigma-Aldrich
氯化钠, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
甲酸, reagent grade, ≥95%
Sigma-Aldrich
氯化钠, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
氯化钠 溶液, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
氯化钠 溶液, 0.9% in water, BioXtra, suitable for cell culture
SAFC
氯化钠 溶液, 5 M
Sigma-Aldrich
氯化钠, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
甲酸, ACS reagent, ≥96%
Sigma-Aldrich
氯化钠 溶液, 5 M
Sigma-Aldrich
甲酸, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥98%
Sigma-Aldrich
乙酸铵, for molecular biology, ≥98%
Sigma-Aldrich
甲醇, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)
Sigma-Aldrich
乙酸铵, reagent grade, ≥98%
Sigma-Aldrich
乙酸铵, ACS reagent, ≥97%
Sigma-Aldrich
四硼酸钠, 99%
Sigma-Aldrich
乙腈, ACS reagent, ≥99.5%
Sigma-Aldrich
乙酸铵, ≥99.99% trace metals basis
Sigma-Aldrich
氯化钠, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
甲醇, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
乙腈 溶液, contains 0.1 % (v/v) formic acid, suitable for HPLC
Sigma-Aldrich
甲酸, puriss., meets analytical specifications of DAC, FCC, 98.0-100%
Sigma-Aldrich
氯化钠 溶液, BioUltra, for molecular biology, ~5 M in H2O
Sigma-Aldrich
氯化钠, BioUltra, for molecular biology, ≥99.5% (AT)
Sigma-Aldrich
乙酸铵 溶液, for molecular biology, 7.5 M
Sigma-Aldrich
乙腈 溶液, contains 0.1 % (v/v) trifluoroacetic acid, suitable for HPLC
Sigma-Aldrich
甲醇, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
乙腈, HPLC Plus, ≥99.9%, poly-coated bottles