On the Origin and Underappreciated Effects of Ion Doping in Silica.
Small (Weinheim an der Bergstrasse, Germany) (2015-06-13)
Xiaohui Song, Tao Ding, Lin Yao, Ming Lin, Rachel Lee Siew Tan, Cuicui Liu, Katarzyna Sokol, Le Yu, Xiong Wen David Lou, Hongyu Chen
摘要
The origin of selectivity in the hollowing of silica nanoparticles is investigated to further understand silica. It is realized that, during the synthesis, the silica precursors are essentially ion-paired polyelectrolytes, whose nucleation depends on the concentration of the counter ions, and most importantly, the size/length of the poly(silicic acid). Thus, the "silica" that nucleates out at the different stages of synthesis has different degrees of ion doping, which explains its solubility in water, its microporosity, and the selective etching phenomena. The etching of silica in water is shown to be a matter of silica solubility, which correlates to the relative amounts of solvent and to the solvent quality (the water/isopropanol ratio). Hollowing does not occur when the silica nanoparticles are incubated in solutions presaturated with "silica," ruling out surface reposition and Ostwald ripening as the hollowing mechanism. The embedded ions in silica are confirmed by elemental analysis (CHNS) and inductively coupled plasma-mass spectrometry. The high ionic doping ratios (N/Si = 2.3% for NH3 -catalyzed silica; Na/Si = 11.2% for NaOH-catalyzed silica) explain the unusual solubility of silica in neutral water. The new view of silica with the ionic impurities on the central stage allows for insights in silica properties and versatility in synthetic design.
材料
货号
品牌
产品描述
Sigma-Aldrich
2-丙醇, BioReagent, for molecular biology, ≥99.5%
Sigma-Aldrich
2-丙醇, anhydrous, 99.5%
Sigma-Aldrich
氢氧化钠 溶液, BioUltra, for molecular biology, 10 M in H2O
Sigma-Aldrich
氢氧化钠 溶液, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
氯化钾, for molecular biology, ≥99.0%
Sigma-Aldrich
亚甲蓝, certified by the Biological Stain Commission
Sigma-Aldrich
异丙醇, ≥99.7%, FCC, FG
Sigma-Aldrich
四甲基氢氧化铵 溶液, 25 wt. % in H2O
Sigma-Aldrich
氯化钾, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99.0%
Sigma-Aldrich
氯化钾, BioXtra, ≥99.0%
Sigma-Aldrich
2-丙醇, BioUltra, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
顺铂, crystalline
Sigma-Aldrich
氰化钾, BioUltra, ≥98.0% (AT)
Sigma-Aldrich
氯化钾 溶液, BioUltra, for molecular biology, ~1 M in H2O
Sigma-Aldrich
11-巯基十一烷酸, 95%
Sigma-Aldrich
苯乙酸, 99%
Sigma-Aldrich
亚甲蓝 溶液, for microscopy, concentrate according to Ehrlich, concentrated, aqueous solution
Sigma-Aldrich
四甲基氢氧化铵 溶液, 25 wt. % in methanol
Sigma-Aldrich
4-巯基苯甲酸, 99%
Sigma-Aldrich
四甲基氢氧化铵 溶液, 10 wt. % in H2O
Sigma-Aldrich
氯化钾 溶液, 0.075 M, sterile-filtered, BioXtra, suitable for cell culture
Sigma-Aldrich
2-丙醇, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
四甲基氢氧化铵 溶液, ACS reagent
Sigma-Aldrich
亚甲蓝 溶液, 0.05 wt. % in H2O
Sigma-Aldrich
氯化钾, 99.999% trace metals basis
Sigma-Aldrich
二氯化二胺(II), ≥99.9% trace metals basis
Sigma-Aldrich
11-巯基十一烷酸, 98%
Sigma-Aldrich
氢氧化钠, BioUltra, for luminescence, ≥98.0% (T), pellets
Sigma-Aldrich
氯化钾, BioUltra, for molecular biology, ≥99.5% (AT)
Sigma-Aldrich
氢氧化钠, ultra dry, powder or crystals, 99.99% trace metals basis