Merck
  • 主页
  • 查找结果
  • Micropatch-arrayed pads for non-invasive spatial and temporal profiling of topical drugs on skin surface.
Micropatch-arrayed pads for non-invasive spatial and temporal profiling of topical drugs on skin surface.
Journal of mass spectrometry : JMS (2015-10-28)
Ewelina P Dutkiewicz, Hsien-Yi Chiu, Pawel L Urban
摘要
Micropatch-arrayed pads (MAPAs) are presented as a facile and sensitive sampling method for spatial profiling of topical agents adsorbed on the surface of skin. MAPAs are 28 × 28 mm sized pieces of polytetrafluoroethylene containing plurality of cavities filled with agarose hydrogel. They are affixed onto skin for 10 min with the purpose to collect drugs applied topically. Polar compounds are absorbed by the hydrogel micropatches. The probes are subsequently scanned by an automated nanospray desorption electrospray ionization mass spectrometry system operated in the tapping dual-polarity mode. When the liquid junction gets into contact with every micropatch, polar compounds absorbed in the hydrogel matrix are desorbed and transferred to the ion source. A 3D-printed interface prevents evaporation of hydrogel micropatches assuring good reproducibility and sensitivity. MAPAs have been applied to follow dispersion of topical drugs applied to human skin in vivo and to porcine skin ex vivo, in the form of self-adhesive patches. Spatiotemporal characteristics of the drug dispersion process have been revealed using this non-invasive test. Differences between drug dispersion in vivo and ex vivo could be observed. We envision that MAPAs can be used to investigate spatiotemporal kinetics of various topical agents utilized in medical treatment.
材料
货号
品牌
产品描述
Sigma-Aldrich
乙腈, anhydrous, 99.8%
Sigma-Aldrich
氢氧化铵 溶液, ACS reagent, 28.0-30.0% NH3 basis
Sigma-Aldrich
氢氧化铵 溶液, 28% NH3 in H2O, ≥99.99% trace metals basis
Sigma-Aldrich
乙腈, ACS reagent, ≥99.5%
Sigma-Aldrich
乙腈 溶液, contains 0.1 % (v/v) trifluoroacetic acid, suitable for HPLC
Sigma-Aldrich
氢氧化铵 溶液, puriss., 30-33% NH3 in H2O
Sigma-Aldrich
乙腈 溶液, contains 0.1 % (v/v) formic acid, suitable for HPLC
Sigma-Aldrich
氢氧化铵 溶液, puriss. p.a., reag. ISO, reag. Ph. Eur., ~25% NH3 basis
Sigma-Aldrich
乙腈, HPLC Plus, ≥99.9%, poly-coated bottles
Sigma-Aldrich
乙腈, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99.5% (GC)
Sigma-Aldrich
氢氧化铵 溶液, puriss., meets analytical specification of Ph. Eur., 25-30% NH3 basis
Sigma-Aldrich
乙腈, biotech. grade, ≥99.93%
Sigma-Aldrich
乙腈, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
乙腈, ReagentPlus®, 99%
Sigma-Aldrich
氢氧化铵 溶液, puriss. p.a. plus, ≥25% NH3 in H2O
Sigma-Aldrich
乙腈, ≥99.5%, ACS reagent
Sigma-Aldrich
乙腈, suitable for DNA synthesis, ≥99.9% (GC)
Sigma-Aldrich
乙腈 溶液, contains 0.05 % (v/v) trifluoroacetic acid
Sigma-Aldrich
乙腈 溶液, contains 10.0% acetone, 40.0% 2-propanol, 0.05% formic acid
Sigma-Aldrich
乙腈 溶液, contains 0.035 % (v/v) trifluoroacetic acid, suitable for HPLC
Sigma-Aldrich
乙腈
Sigma-Aldrich
乙腈 溶液, contains 0.05 % (w/v) ammonium formate, 0.1 % (v/v) formic acid, 5 % (v/v) water, suitable for HPLC
Supelco
乙腈, HPLC grade, ≥99.93%