  • Search Results
  • DanHong injection dose-dependently varies amino acid metabolites and metabolic pathways in the treatment of rats with cerebral ischemia.
DanHong injection dose-dependently varies amino acid metabolites and metabolic pathways in the treatment of rats with cerebral ischemia.
Acta pharmacologica Sinica (2015-05-06)
Zhi-li Guo, Yan Zhu, Xiao-tao Su, Jun Liu, Qian-xu Yang, Jing-yi Nan, Bu-chang Zhao, Ying-ying Zhang, Ya-nan Yu, Bing Li, Hong-bin Xiao, Zhong Wang
ABSTRACT
To determine how the relative amino acid contents and metabolic pathways regulate the pharmacological phenotypes in rats with cerebral ischemia after treatment with varying doses of DanHong injection (DHI). Adult male rats underwent middle cerebral artery occlusion (MCAO), and were injected with DHI (DH-1: 1 mL/kg; DH-2: 2.5 mL/kg; DH-3: 5 mL/kg, and DH-4: 10 mL/kg, iv) daily for 3 d. The neurological deficit score, body weights and infarct volume were assessed. Serum levels of 20 free amino acids were determined using HPLC, and the values were transformed through the quantitative analysis of the amino acids in the serum metabolic spectrum. Multivariate statistical analysis methods (PCA and PLS-DA) and web-based metabolomics tools (MetPa and MetaboAnalyst) were used to analyze the biological data sets for the amino acids. Administration of DHI dose-dependently decreased cerebral infarct volume, and ameliorated neurological deficits. A total of 5, 6, 7 and 7 non-overlapping metabolites were identified in the DH-1, DH-2, DH-3, and DH-4 groups, respectively. Eight metabolites were shared between the DHI groups and the vehicle group. In addition, the serum levels of glutamic acid, aspartic acid and serine increased with increasing DHI dose. A total of 3, 2, 2 and 5 non-overlapping metabolic pathways were identified in the DH-1, DH-2, DH-3 and DH-4 groups, respectively, and glycine, serine, threonine and histidine metabolism were identified as overlapping pathways among the 4 dose groups. Overlapping and non-overlapping amino acid metabolites and metabolic pathways are associated with the dose-dependent neuroprotective effect of DHI.
Sigma-Aldrich
乙腈, anhydrous, 99.8%
Sigma-Aldrich
甲醇, anhydrous, 99.8%
Sigma-Aldrich
邻苯二甲醛, ≥97% (HPLC), powder (may contain lumps)
Sigma-Aldrich
乙腈 溶液, contains 0.1 % (v/v) trifluoroacetic acid, suitable for HPLC
Sigma-Aldrich
乙腈 溶液, contains 0.1 % (v/v) formic acid, suitable for HPLC
Sigma-Aldrich
邻苯二甲醛, suitable for HPLC fluorimetric detection of amino acids, ≥99% (HPLC), powder
Sigma-Aldrich
邻苯二甲醛, for fluorescence, ≥99.0% (HPLC)
Sigma-Aldrich
乙腈, HPLC Plus, ≥99.9%, poly-coated bottles
Sigma-Aldrich
甲醇, HPLC Plus, ≥99.9%, poly-coated bottles
Sigma-Aldrich
辛基磷酸, 97%
Sigma-Aldrich
乙腈, electronic grade, 99.999% trace metals basis
Supelco
甲醇 溶液, contains 0.10 % (v/v) formic acid, UHPLC, suitable for mass spectrometry (MS), ≥99.5%
Supelco
甲醇 溶液, NMR reference standard, 4% in methanol-d4 (99.8 atom % D), NMR tube size 5 mm × 8 in.
Sigma-Aldrich
乙腈, ≥99.5%, ACS reagent
Sigma-Aldrich
甲醇, NMR reference standard
Sigma-Aldrich
乙腈 溶液, contains 0.05 % (v/v) trifluoroacetic acid
Sigma-Aldrich
乙腈 溶液, contains 10.0% acetone, 40.0% 2-propanol, 0.05% formic acid
Sigma-Aldrich
乙腈
Sigma-Aldrich
甲醇 溶液, NMR reference standard, 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.
Sigma-Aldrich
乙腈 溶液, contains 0.05 % (w/v) ammonium formate, 0.1 % (v/v) formic acid, 5 % (v/v) water, suitable for HPLC
Sigma-Aldrich
甲醇-12C, 99.95 atom % 12C
Supelco
乙腈, HPLC grade, ≥99.93%
Sigma-Aldrich
甲醇, suitable for HPLC, gradient grade, 99.93%