• 主页
  • 查找结果
  • Antibody/receptor protein immunocomplex in human and mouse cortical nerve endings amplifies complement-induced glutamate release.
Antibody/receptor protein immunocomplex in human and mouse cortical nerve endings amplifies complement-induced glutamate release.
Neuroscience letters (2015-06-07)
Elisa Merega, Silvia Di Prisco, Paolo Severi, Fotios Kalfas, Anna Pittaluga
摘要
Previous studies have demonstrated that complement alone releases glutamate from human and mouse cortical terminals in an antibody-independent manner. In order to expand our knowledge on complement-mediated effects, we investigated whether the presence of an antigen-antibody complex in synaptosomal plasmamembranes could also trigger complement-induced functional responses that might affect neurotransmitter release. To this aim, we focused on the chemokine 5 receptor (CCR5) expressed in human and mouse cortical glutamate terminals, whose activation by CCL5 elicits [(3)H]D-aspartate ([(3)H]D-ASP) release. Preincubating synaptosomes with an antibody recognizing the NH2 terminus of the CCR5 protein (anti-NH2-CCR5 antibody) abolished the CCL5-induced [(3)H]D-ASP release. Similarly, enriching synaptosomes with an antibody recognizing the COOH terminus of CCR5 (anti-COOH-CCR5 antibody) prevented the CCL5-induced [(3)H]D-ASP release. The antagonist-like activity of the anti-NH2-CCR5 antibody turned to facilitation when anti-NH2-CCR5-treated synaptosomes were exposed to complement. In these terminals, the releasing effect was significantly higher than that elicited by complement in untreated synaptosomes. On the contrary, the complement-induced [(3)H]D-ASP release from anti-COOH-CCR5 antibody-entrapped synaptosomes did not differ from that from untreated synaptosomes. Preincubating synaptosomes with anti-beta tubulin III antibody, used as negative control, neither prevented the CCL5-induced releasing effect nor it amplified the complement-induced [(3)H]D-ASP release. Finally, serum lacking the C1q protein, i.e. the protein essential to promote the antibody-mediated activation of complement, elicited a comparable [(3)H]D-ASP release from both untreated and anti-NH2-CCR5 antibody-treated synaptosomes. Thus, we propose that antibodies raised against the outer sequence of a receptor protein can trigger the activation of the complement through the classic, C1q-mediated antibody-dependent pathway, which results in an abnormal release of glutamate that could be deleterious to central nervous system.
材料
货号
品牌
产品描述
Sigma-Aldrich
羟乙基哌嗪乙硫磺酸, ≥99.5% (titration)
Sigma-Aldrich
碳酸氢钠, powder, BioReagent, for molecular biology, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
羟乙基哌嗪乙硫磺酸, BioPerformance Certified, ≥99.5% (titration), suitable for cell culture
Sigma-Aldrich
氯化钙 溶液, BioUltra, for molecular biology, ~1 M in H2O
Sigma-Aldrich
HEPES缓冲溶液, 1 M in H2O
Sigma-Aldrich
碳酸氢钠, BioXtra, 99.5-100.5%
Sigma-Aldrich
氯化钙, meets USP testing specifications
Sigma-Aldrich
氯化钙, anhydrous, BioReagent, suitable for insect cell culture, suitable for plant cell culture, ≥96.0%
Sigma-Aldrich
氯化钙, anhydrous, powder, 99.99% trace metals basis
SAFC
羟乙基哌嗪乙硫磺酸
Sigma-Aldrich
硫酸镁, BioReagent, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
羟乙基哌嗪乙硫磺酸, BioUltra, for molecular biology, ≥99.5% (T)
Sigma-Aldrich
硫酸镁 溶液, for molecular biology, 1.00 M±0.04 M
SAFC
羟乙基哌嗪乙硫磺酸
Sigma-Aldrich
羟乙基哌嗪乙硫磺酸, BioXtra, pH 5.0-6.5 (1 M in H2O), ≥99.5% (titration)
Sigma-Aldrich
氯化钙, AnhydroBeads, −10 mesh, ≥99.9% trace metals basis
Sigma-Aldrich
硫酸镁 溶液, BioUltra, for molecular biology
Sigma-Aldrich
羟乙基哌嗪乙硫磺酸, BioXtra, suitable for mouse embryo cell culture, ≥99.5% (titration)
Sigma-Aldrich
碳酸氢钠, Hybri-Max, powder, suitable for hybridoma, ≥99.5%
Sigma-Aldrich
羟乙基哌嗪乙硫磺酸, anhydrous, free-flowing, Redi-Dri, ≥99.5%
Sigma-Aldrich
硫酸镁, ≥99.99% trace metals basis
Sigma-Aldrich
氯化钙, AnhydroBeads, −10 mesh, ≥99.99% trace metals basis
Sigma-Aldrich
重碳酸钠-12C, 99.9 atom % 12C