  • Search Results
  • Reorganization of supramammillary-hippocampal pathways in the rat pilocarpine model of temporal lobe epilepsy: evidence for axon terminal sprouting.
Reorganization of supramammillary-hippocampal pathways in the rat pilocarpine model of temporal lobe epilepsy: evidence for axon terminal sprouting.
Brain structure & function (2014-06-04)
Rabia Soussi, Jean-Luc Boulland, Emilie Bassot, Hélène Bras, Patrice Coulon, Farrukh Abbas Chaudhry, Jon Storm-Mathisen, Lotfi Ferhat, Monique Esclapez
ABSTRACT
In mesial temporal lobe epilepsy (MTLE), spontaneous seizures likely originate from a multi-structural epileptogenic zone, including several regions of the limbic system connected to the hippocampal formation. In this study, we investigate the structural connectivity between the supramammillary nucleus (SuM) and the dentate gyrus (DG) in the model of MTLE induced by pilocarpine in the rat. This hypothalamic nucleus, which provides major extracortical projections to the hippocampal formation, plays a key role in the regulation of several hippocampus-dependent activities, including theta rhythms, memory function and emotional behavior, such as stress and anxiety, functions that are known to be altered in MTLE. Our findings demonstrate a marked reorganization of DG afferents originating from the SuM in pilocarpine-treated rats. This reorganization, which starts during the latent period, is massive when animals become epileptic and continue to evolve during epilepsy. It is characterized by an aberrant distribution and an increased number of axon terminals from neurons of both lateral and medial regions of the SuM, invading the entire inner molecular layer of the DG. This reorganization, which reflects an axon terminal sprouting from SuM neurons, could contribute to trigger spontaneous seizures within an altered hippocampal intrinsic circuitry.
Sigma-Aldrich
水, Nuclease-Free Water, for Molecular Biology
Sigma-Aldrich
Triton X-100, laboratory grade
Sigma-Aldrich
过氧化氢 溶液, 30 % (w/w) in H2O, contains stabilizer
Sigma-Aldrich
水, sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
盐酸, 36.5-38.0%, BioReagent, for molecular biology
Sigma-Aldrich
盐酸 溶液, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
水, for molecular biology, sterile filtered
Sigma-Aldrich
水, for embryo transfer, sterile-filtered, BioXtra, suitable for mouse embryo cell culture
Supelco
盐酸 溶液, volumetric, 0.1 M HCl (0.1N), endotoxin free
Sigma-Aldrich
过氧化氢 溶液, 34.5-36.5%
Sigma-Aldrich
钾, chunks (in mineral oil), 98% trace metals basis
Sigma-Aldrich
氯化氢, ReagentPlus®, ≥99%
Sigma-Aldrich
水, BioPerformance Certified
Sigma-Aldrich
水, for cell biology, sterile ultrafiltered
Sigma-Aldrich
氢化钾, 30 wt % dispersion in mineral oil
Supelco
地西泮标准液 溶液, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Sigma-Aldrich
毛果芸香碱 盐酸盐, ≥99% (titration), powder
Sigma-Aldrich
盐酸 溶液, ~6 M in H2O, for amino acid analysis
Sigma-Aldrich
地西泮
Sigma-Aldrich
盐酸 溶液, 32 wt. % in H2O, FCC
Sigma-Aldrich
水, PCR Reagent
Sigma-Aldrich
氯醛 水合物
Sigma-Aldrich
硫代硫酸钠, ≥99.99% trace metals basis
Sigma-Aldrich
氯化氢 溶液, 3 M in cyclopentyl methyl ether (CPME)
Sigma-Aldrich
钾, cubes (in mineral oil), 99.5% trace metals basis
Sigma-Aldrich
E-Toxate 水, endotoxin, free
Sigma-Aldrich
柠檬酸盐浓缩液, BioReagent, suitable for coagulation assays, 4 % (w/v)
Sigma-Aldrich
过氧化氢 溶液, contains potassium stannate as inhibitor, 30-32 wt. % in water, semiconductor grade, 99.999% trace metals basis
Sigma-Aldrich
硫代硫酸钠 溶液, 2 g/dL in deionized water
Sigma-Aldrich
低氘水, ≤1 ppm (Deuterium oxide)