• 主页
  • 查找结果
  • Composite silica coated gold nanosphere and quantum dots nanoparticles for X-ray CT and fluorescence bimodal imaging.
Composite silica coated gold nanosphere and quantum dots nanoparticles for X-ray CT and fluorescence bimodal imaging.
Dalton transactions (Cambridge, England : 2003) (2015-05-27)
Ji-Tao Song, Xiao-Quan Yang, Xiao-Shuai Zhang, Dong-Mei Yan, Ming-Hao Yao, Meng-Yao Qin, Yuan-Di Zhao
摘要
In this study, silica coated Au nanospheres (Au@SiO2) were prepared by a reverse microemulsion method; subsequently, a layer of fluorescent quantum dots (QDs) were adsorbed onto it and then it was coated with silica again. After modifying with PVP, the composite silica coated gold nanosphere and quantum dots nanoparticle (Au@SiO2-QDs/SiO2-PVP) was obtained. This composite structure contained Au and QDs, and it could be used for contrast-enhanced X-ray CT imaging and fluorescence imaging. Characterization showed that the composite nanoparticle had good dispersity, a high fluorescence intensity and a good effect of X-ray absorption, and it was suitable for using as a bimodal imaging probe.
材料
货号
品牌
产品描述
Sigma-Aldrich
纯乙醇, 200 proof, anhydrous, ≥99.5%
Sigma-Aldrich
甲醇, anhydrous, 99.8%
Sigma-Aldrich
纯乙醇, 190 proof, meets USP testing specifications
Sigma-Aldrich
纯乙醇, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
乙醇, anhydrous, denatured
Sigma-Aldrich
甲醇, HPLC Plus, ≥99.9%, poly-coated bottles
Sigma-Aldrich
甲醇, suitable for HPLC, gradient grade, 99.93%
Sigma-Aldrich
DAPI, for nucleic acid staining
Sigma-Aldrich
(3-氨基丙基)三乙氧基硅烷, 99%
Sigma-Aldrich
环己烷, anhydrous, 99.5%
Sigma-Aldrich
(3-氨基丙基)三甲氧基硅烷, 97%
Sigma-Aldrich
(3-氨基丙基)三乙氧基硅烷, ≥98%
Sigma-Aldrich
氨, anhydrous, ≥99.98%
Sigma-Aldrich
硒, powder, −100 mesh, 99.99% trace metals basis
Sigma-Aldrich
羟胺 溶液, 50 wt. % in H2O
Sigma-Aldrich
三正辛基膦, 97%
Sigma-Aldrich
硒, powder, −100 mesh, ≥99.5% trace metals basis
Sigma-Aldrich
正己醇, anhydrous, ≥99%
Sigma-Aldrich
三辛基氧化膦, ReagentPlus®, 99%
Sigma-Aldrich
三正辛基膦, technical grade, 90%
Sigma-Aldrich
十六胺, 98%
Sigma-Aldrich
(3-氨基丙基)三乙氧基硅烷, ≥98.0%
Sigma-Aldrich
氨 溶液, 0.4 M in THF
Sigma-Aldrich
羟胺 溶液, 50 wt. % in H2O, 99.999%
Sigma-Aldrich
六甲基二硅噻吩, synthesis grade
Sigma-Aldrich
氨 溶液, 4 M in methanol
Sigma-Aldrich
三辛基氧化膦, technical grade, 90%
Supelco
10% (v/v) 乙醇标准品, 10 % (v/v) in H2O, analytical standard
Sigma-Aldrich
硒, pellets, <5 mm, ≥99.99% trace metals basis
Sigma-Aldrich
硒, pellets, <5 mm particle size, ≥99.999% trace metals basis