  • Search Results
  • Identification and functional characterization of grapevine transporters that mediate glucose-6-phosphate uptake into plastids.
Identification and functional characterization of grapevine transporters that mediate glucose-6-phosphate uptake into plastids.
Planta (2015-05-27)
Henrique Noronha, Carlos Conde, Serge Delrot, Hernâni Gerós
ABSTRACT
Two grapevine glucose-6-Pi plastidial transporters differently expressed in plant organs and in response to environmental and hormonal signals are characterized. They are involved in starch accumulation in berries and canes. In grapevine, starch accumulation in the trunk is important for winter storage of carbon and in the flower for reproductive development. Berries also accumulate starch in their plastids, which are also involved in the synthesis of aroma compounds important for fruit quality. The present work characterizes two glucose-phosphate translocators (VvGPT1, VvGPT2) that control the accumulation of starch in grape amyloplasts. Three different splicing variants identified for VvGPT2 (VvGPT2α, VvGPT2β and VvGPT2Ω) were more expressed in the leaves than in other organs. In contrast, VvGPT1 transcripts were more abundant in mature berries, canes and flowers than in the leaves. Expression of 35S-VvGPT1-GFP and 35S-VvGPT2Ω-GFP in tobacco leaf epidermal cells showed that the fusion proteins localized at the plastidial envelope. Complementation of the Arabidopsis pgi1-1 mutant impaired in leaf starch synthesis restored its ability to synthesize starch, demonstrating that VvGPT1 and VvGPT2Ω mediate the transport of glucose-6-Pi across the plastidial envelope. In grape cell suspensions, ABA, light and galactinol, together with sucrose and fructose, significantly increased the transcript abundance of VvGPT1, whereas VvGPT2Ω expression was affected only by sucrose. In addition, elicitation with methyl jasmonate strongly upregulated VvGPT1, VvGPT2Ω and VvPAL1, suggesting a role for GPTs in the production of secondary compounds in grapevine. Moreover, in grapevines cultivated in field conditions, VvGPT1 expression was higher in berries more exposed to the sun and subjected to higher temperatures. Although both VvGPT1 and VvGPT2 mediate the same function at the molecular level, they exhibit different expression levels and regulation in plant organs and in response to environmental and hormonal signals.
Sigma-Aldrich
蔗糖, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
D -(+)-葡萄糖, ≥99.5% (GC)
Sigma-Aldrich
蔗糖, ≥99.5% (GC)
Sigma-Aldrich
D -(+)-葡萄糖, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.5%
Sigma-Aldrich
蔗糖, BioXtra, ≥99.5% (GC)
Sigma-Aldrich
溴化十六烷基三甲铵, ≥98%
Sigma-Aldrich
葡萄糖, meets EP, BP, JP, USP testing specifications, anhydrous
Sigma-Aldrich
蔗糖, BioUltra, for molecular biology, ≥99.5% (HPLC)
Sigma-Aldrich
溴化十六烷基三甲铵, for molecular biology, ≥99%
Sigma-Aldrich
D -(+)-葡萄糖, ACS reagent
Sigma-Aldrich
蔗糖, puriss., meets analytical specification of Ph. Eur., BP, NF
Sigma-Aldrich
D -(+)-葡萄糖, BioXtra, ≥99.5% (GC)
Sigma-Aldrich
蔗糖, ACS reagent
Sigma-Aldrich
蔗糖, ≥99.5%
Sigma-Aldrich
蔗糖, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99.5% (GC)
Sigma-Aldrich
溴化十六烷基三甲铵, BioXtra, ≥99%
Sigma-Aldrich
D -(+)-葡萄糖, BioUltra, anhydrous, ≥99.5% (sum of enantiomers, HPLC)
Sigma-Aldrich
溴化十六烷基三甲铵, BioUltra, for molecular biology, ≥99.0% (AT)
Sigma-Aldrich
蔗糖, Grade II, suitable for plant cell culture
Sigma-Aldrich
蔗糖, meets USP testing specifications
Sigma-Aldrich
蔗糖, Grade I, suitable for plant cell culture
Sigma-Aldrich
D -(+)-葡萄糖, suitable for mouse embryo cell culture, ≥99.5% (GC)
Sigma-Aldrich
D -(+)-葡萄糖, Hybri-Max, powder, BioReagent, suitable for hybridoma
SAFC
溴化十六烷基三甲铵
Sigma-Aldrich
D -(+)-葡萄糖, 99.9 atom % 16O, 99.9 atom % 12C