• 主页
  • 查找结果
  • Removal of acetic acid from simulated hemicellulosic hydrolysates by emulsion liquid membrane with organophosphorus extractants.
Removal of acetic acid from simulated hemicellulosic hydrolysates by emulsion liquid membrane with organophosphorus extractants.
Bioresource technology (2015-06-10)
Sang Cheol Lee
摘要
Selective removal of acetic acid from simulated hemicellulosic hydrolysates containing xylose and sulfuric acid was attempted in a batch emulsion liquid membrane (ELM) system with organophosphorus extractants. Various experimental variables were used to develop a more energy-efficient ELM process. Total operation time of an ELM run with a very small quantity of trioctylphosphine oxide as the extractant was reduced to about a third of those required to attain almost the same extraction efficiency as obtained in previous ELM works without any extractant. Under specific conditions, acetic acid was selectively separated with a high degree of extraction and insignificant loss of xylose, and its purity and enrichment ratio in the stripping phase were higher than 92% and 6, respectively. Also, reused organic membrane solutions exhibited the extraction efficiency as high as fresh organic solutions did. These results showed that the current ELM process would be quite practical.
材料
货号
品牌
产品描述
Sigma-Aldrich
纯乙醇, 200 proof, anhydrous, ≥99.5%
Sigma-Aldrich
纯乙醇, 190 proof, meets USP testing specifications
Sigma-Aldrich
纯乙醇, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
乙醇, anhydrous, denatured
Sigma-Aldrich
碳酸钠, BioXtra, ≥99.0%
Sigma-Aldrich
碳酸钠, anhydrous, powder, 99.999% trace metals basis
Sigma-Aldrich
三辛基氧化膦, ReagentPlus®, 99%
Sigma-Aldrich
磷酸三丁酯, ≥99%
Sigma-Aldrich
磷酸三丁酯, 97%
Sigma-Aldrich
三辛基氧化膦, technical grade, 90%
Sigma-Aldrich
碳酸钠, BioUltra, anhydrous, ≥99.5% (calc. on dry substance, T)
Supelco
10% (v/v) 乙醇标准品, 10 % (v/v) in H2O, analytical standard
Sigma-Aldrich
三丁基膦 溶液, 200 mM (in N-methyl-2-pyrrolidinone), liquid
Sigma-Aldrich
80% v/v 乙醇固定液, suitable for fixing solution (blood films)
Sigma-Aldrich
碳酸钠-12C, 99.9 atom % 12C