Merck
  • 主页
  • 查找结果
  • Characterization of lemon (Citrus limon) polar extract by liquid chromatography-tandem mass spectrometry in high resolution mode.
Characterization of lemon (Citrus limon) polar extract by liquid chromatography-tandem mass spectrometry in high resolution mode.
Journal of mass spectrometry : JMS (2015-10-28)
C A Ledesma-Escobar, F Priego-Capote, M D Luque de Castro
摘要
Eighty four metabolites (32 flavonoids, 15 amino acids, nine carboxylic acids, six coumarins, six sugars, five phenolic acids and 11 unclassified compounds) have been tentatively identified in a polar extract from lemon, without reference standards, based on their liquid chromatography-quadrupole-time-of-flight MS/MS spectra and the comparison with databases. Despite information in databases for some families of plant compounds is poor, tentative identification based on MS/MS information (mass of the precursor ion and their fragments, together with neutral mass loss) was possible with the help of known fragmentation patterns for the given families of compounds. Both positive and negative ionization modes and at least two collision energies were always applied to obtain as much information as possible from each molecular entity, thus helping for identification. As the tentatively identified metabolites are the same regardless of the organism they belong, their fragmentation patterns are useful for identification with independence of the sample nature.
材料
货号
品牌
产品描述
Sigma-Aldrich
纯乙醇, 200 proof, for molecular biology
Sigma-Aldrich
乙腈, anhydrous, 99.8%
Sigma-Aldrich
纯乙醇, 200 proof, anhydrous, ≥99.5%
Sigma-Aldrich
甲醇, anhydrous, 99.8%
Sigma-Aldrich
纯乙醇, 200 proof, ACS reagent, ≥99.5%
Sigma-Aldrich
甲醇, ACS reagent, ≥99.8%
Sigma-Aldrich
甲酸, reagent grade, ≥95%
Sigma-Aldrich
纯乙醇, 200 proof, HPLC/spectrophotometric grade
Sigma-Aldrich
甲酸, ACS reagent, ≥96%
Sigma-Aldrich
乙腈, ACS reagent, ≥99.5%
Sigma-Aldrich
甲酸, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥98%
Sigma-Aldrich
纯乙醇, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
纯乙醇, 190 proof, for molecular biology
Sigma-Aldrich
纯乙醇, 190 proof, meets USP testing specifications
Sigma-Aldrich
甲醇, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)
Sigma-Aldrich
纯乙醇, 200 proof, meets USP testing specifications
Sigma-Aldrich
甲酸, puriss., meets analytical specifications of DAC, FCC, 98.0-100%
Sigma-Aldrich
乙腈 溶液, contains 0.1 % (v/v) trifluoroacetic acid, suitable for HPLC
Sigma-Aldrich
乙腈 溶液, contains 0.1 % (v/v) formic acid, suitable for HPLC
Sigma-Aldrich
甲醇, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
乙醇, ACS reagent, prima fine spirit, without additive, F15 o1
Sigma-Aldrich
甲酸, ACS reagent, ≥88%
Sigma-Aldrich
甲醇, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
甲醇, BioReagent, ≥99.93%
Sigma-Aldrich
甲醇, Absolute - Acetone free
Sigma-Aldrich
乙腈, HPLC Plus, ≥99.9%, poly-coated bottles
Sigma-Aldrich
甲醇, ACS reagent, ≥99.8%
Sigma-Aldrich
乙腈, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99.5% (GC)
Sigma-Aldrich
甲醇, puriss., meets analytical specification of Ph Eur, ≥99.7% (GC)
Sigma-Aldrich
甲酸, ≥95%, FCC, FG