  • Search Results
  • Fatty acid effect on sucrose-induced enamel demineralization and cariogenicity of an experimental biofilm-caries model.
Fatty acid effect on sucrose-induced enamel demineralization and cariogenicity of an experimental biofilm-caries model.
Odontology (2014-04-12)
Rodrigo A Giacaman, Pascale Jobet-Vila, Cecilia Muñoz-Sandoval
ABSTRACT
Based on scarce evidence, fatty acids have been described as anticariogenic. The aim was to evaluate the effect of different types of fatty acids on enamel demineralization and on the cariogenic properties of Streptococcus mutans biofilms on a biofilm/caries model. Mature biofilms of S. mutans UA159 growing on bovine enamel slabs were exposed to 10% sucrose for 5 min, 3 times per day followed by exposure to a panel of free fatty acids, including monounsaturated (oleic), polyunsaturated (linoleic) and saturated (stearic) fatty acids, in concentrations of 0.1, 1 and 10 mM for five additional minutes. Enamel demineralization was determined before and after the experiments by microhardness. Slabs were retrieved to analyze biofilm biomass, viable bacterial counts and polysaccharide production. Biofilms exposed to sucrose, followed by oleic and linoleic acids, showed less demineralization than sucrose alone (p < 0.05). Biomass, S. mutans colonies and insoluble extracellular polysaccharide production were reduced from the biofilms treated with oleic and linoleic fatty acids (p < 0.05). No differences with the positive control were observed with the saturated stearic acid. Poly and monounsaturated fatty acids presented to S. mutans biofilms after a cariogenic challenge appear to reduce demineralization on enamel and to interfere with cariogenicity of S. mutans biofilms.
Sigma-Aldrich
纯乙醇, 200 proof, anhydrous, ≥99.5%
Sigma-Aldrich
氯化钠, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
蔗糖, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
蔗糖, ≥99.5% (GC)
Sigma-Aldrich
氯化钠, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
蔗糖, BioXtra, ≥99.5% (GC)
Sigma-Aldrich
氯化钠 溶液, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
氯化钠, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
氯化钠 溶液, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
蔗糖, BioUltra, for molecular biology, ≥99.5% (HPLC)
SAFC
氯化钠 溶液, 5 M
Sigma-Aldrich
氯化钠 溶液, 5 M
Sigma-Aldrich
蔗糖, puriss., meets analytical specification of Ph. Eur., BP, NF
Sigma-Aldrich
纯乙醇, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
纯乙醇, 190 proof, meets USP testing specifications
Sigma-Aldrich
氯化钠, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
蔗糖, ACS reagent
Sigma-Aldrich
蔗糖, ≥99.5%
Sigma-Aldrich
蔗糖, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99.5% (GC)
Sigma-Aldrich
氯化钠, BioUltra, for molecular biology, ≥99.5% (AT)
Sigma-Aldrich
氯化钠 溶液, 0.85%
Sigma-Aldrich
氯化钠 溶液, BioUltra, for molecular biology, ~5 M in H2O
Sigma-Aldrich
蔗糖, Grade II, suitable for plant cell culture
Sigma-Aldrich
蔗糖, meets USP testing specifications
Sigma-Aldrich
氯化钠, 99.999% trace metals basis
Sigma-Aldrich
蔗糖, Grade I, suitable for plant cell culture
Sigma-Aldrich
氯化钠, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Sigma-Aldrich
氢碘酸, 57 wt. %, distilled, 99.999% trace metals basis
Supelco
10% (v/v) 乙醇标准品, 10 % (v/v) in H2O, analytical standard
Sigma-Aldrich
氯化钠, tablet